Alignflow™ Flow Cytometry Alignment Beads for Red Lasers, 6.0 μm
Invitrogen™

Alignflow™ Flow Cytometry Alignment Beads for Red Lasers, 6.0 μm

Molecular Probes™ AlignFlow™ flow cytometry alignment beads are reliable references for aligning, focusing, and calibrating flow cytometers.The 6.0 μm Alignflow™자세히 알아보기
Have Questions?
카탈로그 번호수량
A16504
A-16504으로도 사용됨
3 mL
카탈로그 번호 A16504
A-16504으로도 사용됨
제품 가격(KRW)
539,000
온라인 행사
Ends: 30-Sep-2026
633,000
할인액 94,000 (15%)
Each
수량:
3 mL
제품 가격(KRW)
539,000
온라인 행사
Ends: 30-Sep-2026
633,000
할인액 94,000 (15%)
Each
Molecular Probes™ AlignFlow™ flow cytometry alignment beads are reliable references for aligning, focusing, and calibrating flow cytometers.The 6.0 μm Alignflow™ Flow Cytometry Alignment Beads for Red Lasers are fluorescent-dye infused microspheres that are excitable at 630–640 nm with emission in the range of 645–680 nm.

View information for all flow cytometry calibration and size reference beads.

AlignFlow™ beads provide:

• Uniformity—very low FSC CV and intensity CV
• Consistency in production, creating ideal reference standards
• Confidence through reliable, optimal instrument performance
• Compatibility—use with any instrument

The AlignFlow™ beads are highly uniform with respect to size and fluorescence intensity, resulting in low FSC CV and intensity CV. The beads approximate the size, emission wavelength, and intensity of many biological samples and permit the calibration of the flow cytometer’s laser source, optics, and stream flow without wasting valuable and sensitive experimental material.

Bead Labeling
As with all of our flow cytometry standard microspheres, the AlignFlow™ beads are stained internally rather than on the surface. The dyes are therefore insulated from environmental interactions that could cause variable fluorescence output, resulting in excellent signal stability. These fluorescent polystyrene microspheres are supplied as suspensions packaged in dropper vials for convenient dispensation and are available in a choice of three colors (UV, blue, red) and two sizes (2.5 μm, 6.0 μm) to match your laser.

Notes:
The AlignFlow™ flow cytometry reference beads are provided in updated packaging that includes a Quick Reference Label that puts key information for product use at your fingertips.

These beads replace previous AlignFlow™ flow cytometry reference bead products. This particular product replaces AlignFlow™ Plus Flow Cytometry Alignment Beads, 6.0 μm, for 633 excitation (A7313).
For Research Use Only. Not for use in diagnostic procedures.
사양
검출 방법Fluorescence
직경(미터법)6 μm
여기 파장 범위633 or 635
용도(장비)Flow Cytometer
형식Suspension
수량3 mL
배송 조건Room Temperature
염색 구성Core
제품라인AlignFlow
제품 유형Alignment Beads
Unit SizeEach
구성 및 보관
Contains 2 vials of Alignflow™ beads (1.5 mL per vial). Store at 2–8°C.

자주 묻는 질문(FAQ)

What are Alignflow Flow Cytometry Alignment Beads used for?

Alignflow Flow Cytometry Alignment Beads are reliable references for aligning, focusing, and calibrating flow cytometers. The Alignflow beads are highly uniform with respect to size and fluorescence intensity, resulting in low CVs for both FSC and intensity. The beads approximate the size, emission wavelength, and intensity of many biological samples and permit the calibration of the flow cytometer’s laser source, optics, and stream flow without wasting valuable and sensitive experimental material. They are available in 2.5 µM and 6.0 µM sizes for blue, red, and UV lasers. Alignflow Flow Cytometry Alignment Beads for UV Lasers are optimally excitable between 350-370 nm with emission in the range of 400-470 nm, but they can also be excited by a violet laser.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.