Alignflow™ Flow Cytometry Alignment Beads for UV Lasers, 6.0 μm
Invitrogen™

Alignflow™ Flow Cytometry Alignment Beads for UV Lasers, 6.0 μm

Molecular Probes™ AlignFlow™ flow cytometry alignment beads are reliable references for aligning, focusing, and calibrating flow cytometers.The 6.0 μm AlignFlow™Read more
Have Questions?
Catalog NumberQuantity
A16505
also known as A-16505
3 mL
Catalog number A16505
also known as A-16505
Price (USD)
330.65
Online exclusive
441.00
Save 110.35
Each
Quantity:
3 mL
Price (USD)
330.65
Online exclusive
441.00
Save 110.35
Each
Molecular Probes™ AlignFlow™ flow cytometry alignment beads are reliable references for aligning, focusing, and calibrating flow cytometers.The 6.0 μm AlignFlow™ Flow Cytometry Alignment Beads for UV Lasers are fluorescent-dye infused microspheres that are optimally excitable between 350–370 nm with emission in the range of 400–470 nm, but that can also be excited by a violet laser.

View information for all flow cytometry calibration and size reference beads.

AlignFlow™ beads provide:

• Uniformity—very low FSC CV and intensity CV
• Consistency in production, creating ideal reference standards
• Confidence through reliable, optimal instrument performance
• Compatibility—use with any instrument

The AlignFlow™ beads are highly uniform with respect to size and fluorescence intensity, resulting in low FSC CV and intensity CV. The beads approximate the size, emission wavelength, and intensity of many biological samples and permit the calibration of the flow cytometer’s laser source, optics, and stream flow without wasting valuable and sensitive experimental material.

Bead Labeling
As with all of our flow cytometry standard microspheres, the AlignFlow™ beads are stained internally rather than on the surface. The dyes are therefore insulated from environmental interactions that could cause variable fluorescence output, resulting in excellent signal stability. These fluorescent polystyrene microspheres are supplied as suspensions packaged in dropper vials for convenient dispensation and are available in a choice of three colors (UV, blue, red) and two sizes (2.5 μm, 6.0 μm) to match your laser.

Notes:
The AlignFlow™ flow cytometry reference beads are provided in updated packaging that includes a Quick Reference Label that puts key information for product use at your fingertips.

These beads replace previous AlignFlow™ flow cytometry reference bead products. This particular product replaces AlignFlow™ Plus Flow Cytometry Alignment Beads, 6.0 μm, for UV excitation (A7305).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Diameter (Metric)6 μm
EmissionUV
For Use With (Equipment)Flow Cytometer
FormatSuspension
Quantity3 mL
Shipping ConditionRoom Temperature
Stain ConfigurationCore
Product LineAlignFlow
Product TypeAlignment Beads
Unit SizeEach
Contents & Storage
Contains 2 vials of Alignflow™ beads (1.5 mL per vial). Store at 2–8°C.

Frequently asked questions (FAQs)

What are Alignflow Flow Cytometry Alignment Beads used for?

Alignflow Flow Cytometry Alignment Beads are reliable references for aligning, focusing, and calibrating flow cytometers. The Alignflow beads are highly uniform with respect to size and fluorescence intensity, resulting in low CVs for both FSC and intensity. The beads approximate the size, emission wavelength, and intensity of many biological samples and permit the calibration of the flow cytometer’s laser source, optics, and stream flow without wasting valuable and sensitive experimental material. They are available in 2.5 µM and 6.0 µM sizes for blue, red, and UV lasers. Alignflow Flow Cytometry Alignment Beads for UV Lasers are optimally excitable between 350-370 nm with emission in the range of 400-470 nm, but they can also be excited by a violet laser.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.