Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • OxPhos Complex IV subunit IV Antibodies

          Invitrogen

          OxPhos Complex IV subunit IV Monoclonal Antibody (20E8C12)

          56 Published Figures
          135 References
          View all (2) OxPhos Complex IV subunit IV antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite OxPhos Complex IV subunit IV Monoclonal Antibody (20E8C12)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Published Figures (56)
          OxPhos Complex IV subunit IV Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          OxPhos Complex IV subunit IV Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 56

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          OxPhos Complex IV subunit IV Antibody (A21348) in ICC/IF

          PloS one 2012 - Figure 2 Determination of the subcellular localization of ABCB6 by double immunofluorescence labeling and laser-scanning confocal microscopy. A . Expression of the endogenous ABCB6 protein in K562 cells was visualized by the monoclonal ABCB6-567 antibody (green), lysosomes and mitochondria were labeled with LAMP1 (red, lower panel) and CoxIV antibodies (red, upper panel), respectively. ABCB6 colocalizes wit... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          OxPhos Complex IV subunit IV Antibody in Immunocytochemistry (ICC/IF)
          OxPhos Complex IV subunit IV Antibody in Immunocytochemistry (ICC/IF)
          OxPhos Complex IV subunit IV Antibody in Immunocytochemistry (ICC/IF)
          OxPhos Complex IV subunit IV Antibody in Immunocytochemistry (ICC/IF)
          OxPhos Complex IV subunit IV Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          OxPhos Complex IV subunit IV Antibody in Immunohistochemistry (IHC)
          OxPhos Complex IV subunit IV Antibody in Immunohistochemistry (IHC)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)
          OxPhos Complex IV subunit IV Antibody in Western Blot (WB)

          Product Details

          A21348

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1.0 µg/mL
          View 114 publications 114 publications

          Immunohistochemistry (IHC)

          -
          View 7 publications 7 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 6 publications 6 publications

          ELISA (ELISA)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 5 publications 5 publications
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Rat, Zebrafish

          Published species

          Human, Mouse, Non-human primate, Rat, Yeast

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          20E8C12

          Immunogen

          Bovine Complex IV (native holoenzyme protein, purified from liver).

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          purified

          Storage buffer

          HEPES buffered saline

          Contains

          0.02% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2535839

          Target Information

          Complex IV, also called cytochrome c oxidoreductase or cytochrome c oxidase, is a complex of 13 different subunits, three of which (I, II and III) are encoded on mitochondrial DNA and the remainder in the nuclear DNA. The complex contains two heme groups (a and a3) and two copper atoms as prosthetic groups.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline