Albumin from Bovine Serum (BSA), Texas Red™ conjugate
This albumin from bovine serum (BSA), Texas Red™ conjugate is suited for applications in which well-defined molecular weight tracers areWeitere Informationen
Have Questions?
Katalognummer
Menge
A23017
5 mg
Katalognummer A23017
Preis (EUR)
251,65
Exklusiv online
286,00
Ersparnis 34,35 (12%)
Each
Menge:
5 mg
Preis (EUR)
251,65
Exklusiv online
286,00
Ersparnis 34,35 (12%)
Each
This albumin from bovine serum (BSA), Texas Red™ conjugate is suited for applications in which well-defined molecular weight tracers are needed. These applications include studying the rate of endocytosis and exocytosis and following intracellular protein processing.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
MarkertypClassic Dyes
ProduktlinieTexas Red
Protein-SubtypAlbumin Proteins
Menge5 mg
VersandbedingungRoom Temperature
KonjugatTexas Red
FormLyophilized
SpeziesBovine
Unit SizeEach
Inhalt und Lagerung
Store in freezer (-5 to -30°C) and protect from light.
Zitierungen und Referenzen (26)
Zitierungen und Referenzen
Abstract
Nuclear localization and export signals of the human aryl hydrocarbon receptor.
Authors:Ikuta T, Eguchi H, Tachibana T, Yoneda Y, Kawajiri K
Journal:J Biol Chem
PubMed ID:9446600
'The aryl hydrocarbon receptor (Ahr) is a ligand-activated transcription factor that binds DNA in the form of a heterodimer with the Ahr nuclear translocator (hypoxia-inducible factor 1beta). We found in this study that Ahr contains both nuclear localization and export signals in the NH2-terminal region. A fusion protein composed of ... More
Enzymatic activity of alkaline phosphatase inside protein and polymer structures fabricated via multiphoton excitation.
Authors:Basu S, Campagnola PJ
Journal:Biomacromolecules
PubMed ID:15003023
'We demonstrate micron scale control of bioactivity through the use of multiphoton excited photochemistry, where this technique has been used to cross-link three-dimensional matrixes of alkaline phosphatase, bovine serum albumin, and polyacrylamide and combinations therein. Using a fluorescence-based assay (ELF-97), the enzymatic activity has been studied using a Michaelis-Menten analysis, ... More
Regulation of albumin endocytosis by PSD95/Dlg/ZO-1 (PDZ) scaffolds. Interaction of Na+-H+ exchange regulatory factor-2 with ClC-5.
Authors:Hryciw DH, Ekberg J, Ferguson C, Lee A, Wang D, Parton RG, Pollock CA, Yun CC, Poronnik P
Journal:J Biol Chem
PubMed ID:16601121
'The constitutive reuptake of albumin from the glomerular filtrate by receptor-mediated endocytosis is a key function of the renal proximal tubules. Both the Cl- channel ClC-5 and the Na+-H+ exchanger isoform 3 are critical components of the macromolecular endocytic complex that is required for albumin uptake, and therefore the cell-surface ... More
Trichocyst membrane fate in living Paramecium primaurelia visualized by multimodal confocal imaging.
Authors:Ramoino P, Beltrame F, Diaspro A, Fato M
Journal:Eur J Cell Biol
PubMed ID:9309393
'Trichocysts are secretory organelles located at the surface of several ciliates, docked at the plasma membrane. Their secretion is similar to other exocytic processes: the trichocyst membrane fuses with the plasma membrane, its content is released outside the cell, and the membrane is retrieved back into the cell. The fate ... More
High refractive index substrates for fluorescence microscopy of biological interfaces with high z contrast.
Authors:Ajo-Franklin CM, Kam L, Boxer SG
Journal:Proc Natl Acad Sci U S A
PubMed ID:11717428
'Total internal reflection fluorescence microscopy is widely used to confine the excitation of a complex fluorescent sample very close to the material on which it is supported. By working with high refractive index solid supports, it is possible to confine even further the evanescent field, and by varying the angle ... More