SILAC DMEM Flex Media, no glucose, no phenol red
Gibco™

SILAC DMEM Flex Media, no glucose, no phenol red

SILAC DMEM Flex is Dulbecco’s Modified Eagle Medium (DMEM) without L-glutamine, L-lysine, and L-arginine for use during SILAC protein labeling자세히 알아보기
Have Questions?
카탈로그 번호수량
A2493901500 mL
카탈로그 번호 A2493901
제품 가격(KRW)
140,000
온라인 행사
Ends: 30-Sep-2026
164,000
할인액 24,000 (15%)
Each
수량:
500 mL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
140,000
온라인 행사
Ends: 30-Sep-2026
164,000
할인액 24,000 (15%)
Each
SILAC DMEM Flex is Dulbecco’s Modified Eagle Medium (DMEM) without L-glutamine, L-lysine, and L-arginine for use during SILAC protein labeling with stable isotopic labeled lysine and/or arginine. DMEM is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of applications. Find the right modification using the media selector tool.

This DMEM is modified as follows:
WithWithout
 • Glucose
 • Phenol red
 • L-arginine
 • L-glutamine
 • L-lysine
 • Sodium pyruvate
 • HEPES

DMEM is unique from other media as it contains 4 times the concentrations of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. SILAC DMEM Flex has been further modified to not contain lysine, arginine, and glutamine. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). SILAC protein labeling experiments should be conducted using Gibco™ Dialyzed FBS. DMEM uses a sodium bicarbonate buffer system (3.7 g/L), so requires a 5-10% CO2 environment to maintain physiological pH.

cGMP Manufacturing and Quality System
SILAC DMEM Flex is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
사양
세포 유형Mammalian Cells
제품라인Gibco
제품 유형Dulbecco's Modified Eagle Medium
수량500 mL
유통 기한12 Months
Culture TypeMammalian Cell Culture
형태Liquid
Serum LevelStandard Serum Supplementation
첨가제 없음No Glucose, No Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate, No Arginine, No Lysine
Unit SizeEach
구성 및 보관
Store in refrigerator (2–8°C) and protect from light.
Shelf life: 12 months from date of manufacture.

자주 묻는 질문(FAQ)

Do you offer media without phenol red (phenol red-free media)?

Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.