Glucose Solution - FAQs
Glucose Solution - FAQs
View additional product information for Glucose Solution - FAQs (A2494001)
2 product FAQs found
What is the glucose concentration in Glucose Solution (Cat. No. A249001)?
The glucose concentration of Glucose Solution (Cat. No. A2494001) is 200 grams of glucose per 1L of distilled water.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What solvent is used in the Glucose Solution (Cat. No. A2494001)?
The Glucose Solution (Cat. No. A2494001) solvent is distilled water.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.