QuantStudio™ 5 Spectral Calibration Plate 1 (FAM™, VIC™, ROX™, TAMRA™, and SYBR™ dyes), 384-well - FAQs
QuantStudio™ 5 Spectral Calibration Plate 1 (FAM™, VIC™, ROX™, TAMRA™, and SYBR™ dyes), 384-well - FAQs
View additional product information for QuantStudio™ 5 Spectral Calibration Plate 1 (FAM™, VIC™, ROX™, TAMRA™, and SYBR™ dyes), 384-well - FAQs (A26334)
2 product FAQs found
Do I need to perform a normalization calibration for QuantStudio 6 and 7 Flex Real-Time PCR System Software 1.7 after running a QuantStudio 5 Spectral Calibration Plate 1, 384 well (Cat. No. A26334)?
No, you do not need to perform the normalization calibration. The following link, Real-Time PCR Instrument Calibration & Verification Kits, in the “QuantStudio 7 Flex” section, shows that normalization plates are not included if the software is version 1.6 or later.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Is the QuantStudio 5 Spectral Calibration Plate 1, 384 well (Cat. No. A26334) compatible with the QuantStudio 7 Flex Real-Time PCR System, 384 well (Cat. No. 4485701)?
Yes, the QuantStudio 5 Spectral Calibration Plate 1, 384 well is compatible with the QuantStudio 7 Flex Real-Time PCR System, 384 well, with version 1.6, or later, of the corresponding software. However, this plate is not compatible with previous versions of the software.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.