Taking your work to the clinic? Check out CTS™ RevitaCell™ Supplement (100X) »
RevitaCell™ Supplement (100X)
Gibco™

RevitaCell™ Supplement (100X)

RevitaCell™ Supplement (100X) has been optimized for use with pluripotent stem cells (PSCs), either as a post-thaw recovery solution toRead more
Have Questions?
Catalog NumberQuantity
A26445015 mL
Catalog number A2644501
Price (USD)
259.00
Each
Quantity:
5 mL
Price (USD)
259.00
Each
RevitaCell™ Supplement (100X) has been optimized for use with pluripotent stem cells (PSCs), either as a post-thaw recovery solution to improve cell viability or in combination with Essential 8™ Medium to enhance single cell passaging applications. It can also be used for recovery of primary neurons, traditionally a challenging cell to efficiently recover. The significant improvement seen in these applications is due to the presence of a more specific ROCK inhibitor coupled with molecules containing antioxidant and free radical scavenger properties that minimize the impact of stress on the cells.

• Enabling—provides maximum cell viability when performing single cell passaging
• Rapid cell recovery—high post-thaw cell viability and growth of pluripotent stem cells (passaged as clumps or single cells) and neurons
• Consistent—formulated using animal origin-free and chemically defined materials for quality and confidence

Single cell passaging
Use of the RevitaCellTM Supplement for 18-24 hours post-single cell passaging provides vast improvement in cell survival. Cells are shown to reach passaging confluency within 3-4 days post-seeding.

Higher specificity
Kinase profiling of pro-survival small molecules in RevitaCell™ Supplement shows higher specificity in comparison to traditional ROCK inhibitors (i.e., Y-27632 and Thiazovivin).

Quicker post-thaw recovery
PSCs recovered at 40,000 viable cells/cm2 using media supplemented with RevitaCell™ Supplement for the first 24 hours post-thaw recovery followed by feeding with unsupplemented Essential 8™ Medium will typically reach 80% confluency in <5 days, significantly reducing the average post-thaw recovery time of most cryopreservation solutions. Neurite outgrowth of neurons is also significantly improved using RevitaCell™ Supplement.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeStem Cells (iPS - Induced Pluripotent Stem)
Concentration100 X
Culture TypeStem Cell (Embryonic, iPS - Induced Pluripotent Stem)
Manufacturing QualitycGMP for medical devices, 21 CFR Part 820 and ISO 13485
Product LineGibco, RevitaCell
Quantity5 mL
Shipping ConditionDry Ice
SourceAnimal origin-free
ClassificationAnimal Origin-free
Product TypeMedia Supplement
Unit SizeEach
Contents & Storage
Store at -5 to -20°C and protect from light.

Frequently asked questions (FAQs)

Can RevitaCell Supplement be used in addition to a traditional ROCK Inhibitor?

No. Combination of RevitaCell Supplement with a traditional ROCK inhibitor will result in deleterious effects on PSCs.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

The RevitaCell Supplement in the PSC Cryopreservation Kit has a strong smell. Is this normal?

Yes, it is normal that the RevitaCell Supplement has a strong smell.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the difference between the PSC Cryopreservation Kit and the CTS PSC Cyropreservation Kit?

The CTS PSC Cryopreservation Kit (Cat. No. A4239301) provides the same efficient recovery, consistency, and reduced variability as the PSC Cryopreservation Kit (Cat. No. A2644601). However, these two kits have different intended use statements, i.e., “For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals” and “For Research Use Only”, respectively. Furthermore, the CTS PSC Cryopreservation Kit has full USP sterility testing info and a Drug Master file (DMF), whereas the sterility testing of the PSC Cryopreservation is via a modified USP method, and it does not have a DMF.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is StemFlex Medium compatible with downstream differentiation kits available from Thermo Fisher Scientific?

Yes. We have seen compatibility with the following differentiation kits provided by Thermo Fisher Scientific: PSC Cardiomyocyte Differentiation Kit (Cat. No. A2921201), PSC Definitive Endoderm Induction Kit (Cat. No. A3062601), PSC Neural Induction Medium (Cat. No. A1647801), and PSC Dopaminergic Neuron Differentiation Kit (Cat. No. A3147701).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

I see in the flexible feeding schedules specified for StemFlex Medium that cells should be fed the day after passaging, however this is incompatible with my current workflow. Have you evaluated skipping the feed the day after passaging?

Yes. The main reason that the workflow states this is to accommodate ROCK inhibitor or RevitaCell Supplement that have been added during passaging. In such circumstances where ROCK inhibitor or RevitaCell Supplement are added, you should feed your cells within 18-24 hours post-passaging. However, if ROCK inhibitor or RevitaCell Supplement are not included at the time of passaging, then you may skip the day post-passaging.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.