Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Secondary Antibodies ›
          • Goat anti-Rabbit IgG (Heavy chain) Secondary Antibodies
          Discover how to image with Invitrogen F(ab) secondary antibodies

          Invitrogen

          Goat anti-Rabbit IgG (Heavy chain) Superclonal™ Secondary Antibody, Alexa Fluor™ 488

          Advanced Verification
          21 Published Figures
          View all (10) Goat anti-Rabbit IgG (Heavy chain) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Questions & Answers
          Tech Support

          Cite Goat anti-Rabbit IgG (Heavy chain) Superclonal™ Secondary Antibody, Alexa Fluor™ 488

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (25)
          • Published Figures (21)
          • Advanced Verification (1)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 47

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Rabbit IgG (Heavy chain) Secondary Antibody (A27034) in ICC/IF

          Immunofluorescence analysis of Goat anti-Rabbit IgG (Heavy Chain), Superclonal™ Recombinant Secondary Antibody, Alexa Fluor™ 488 (Product # A27034) was performed using MCF 10A (positive model) and A-431 (negative model) in cells stained with Vimentin Polyclonal antibody (Product # PA5-27231). The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Rabbit IgG (Heavy chain) Secondary Antibody in Flow Cytometry (Flow)
          Rabbit IgG (Heavy chain) Secondary Antibody in Flow Cytometry (Flow)
          Rabbit IgG (Heavy chain) Secondary Antibody in Flow Cytometry (Flow)
          Rabbit IgG (Heavy chain) Secondary Antibody in Flow Cytometry (Flow)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Rabbit IgG (Heavy chain) Secondary Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Rabbit IgG (Heavy chain) Secondary Antibody in Western Blot (WB)
          Rabbit IgG (Heavy chain) Secondary Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          A27034

          Applications
          Tested Dilution

          Western Blot (WB)

          1:2,000-1:5,000

          Immunohistochemistry (Frozen) (IHC (F))

          1:2,000

          Immunocytochemistry (ICC/IF)

          1:250-1:2,000

          Flow Cytometry (Flow)

          1-5 µg/mL
          Product Specifications

          Species Reactivity

          Rabbit

          Host/Isotype

          Goat / IgG

          Expression System

          Expi293

          Class

          Recombinant Superclonal

          Type

          Secondary Antibody

          Immunogen

          Recombinant full-length protein

          Conjugate

          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488
          • Alexa Fluor 555 
          • Alexa Fluor 647 
          • Alexa Fluor 790 
          • Alexa Fluor Plus 405 
          • Alexa Fluor Plus 488 
          • Alexa Fluor Plus 647 
          • Biotin 
          • HRP
          • Unconjugated
          • Request custom conjugation

          Excitation/Emission Max

          499/520 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          1 mg

          Purification

          Gravity column chromatography

          Storage buffer

          PBS, pH 7.2-7.4

          Contains

          5mM sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Wet ice

          RRID

          AB_2536097

          Target

          IgG (Heavy chain)

          Antibody Form

          Whole Antibody

          Product Specific Information

          The sensitivity and specificity of each lot is confirmed using ELISA.



          Minimal cross-reactivity with mouse, rat, human, bovine, guinea pig and donkey IgG is observed.



          Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.

          Target Information

          Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          View product citations for antibody A27034 on CiteAb

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0