ProFlex™ PCR System Extended Warranty Package, 96-well
Applied Biosystems™
ProFlex™ PCR System Extended Warranty Package, 96-well
ProFlex™ PCR Systems are the latest in our line of Applied Biosystems™ thermal cyclers, providing the reliability and performance you’ve come to expect from Applied Biosystems instruments. This package includes the ProFlex PCR System and an extended warranty.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWarranty
A279353 years of total service coverage with instrument rapid exchange
A279345 years of total service coverage with instrument rapid exchange
A279365 years of total service coverage with 3 PMs
Catalog number A27935
Price (USD)
14,191.93
Each
Warranty:
3 years of total service coverage with instrument rapid exchange
Price (USD)
14,191.93
Each
ProFlex™ PCR Systems are the latest in our line of Applied Biosystems™ thermal cyclers, providing the reliability and performance you've come to expect from Applied Biosystems instruments. By combining an extended warranty with your instrument purchase, you'll extend the life of your investment and receive the support you need to keep your instrument running at peak performance.
Each ProFlex PCR System Extended Warranty Package includes instrument configuration and control features, as well as extended warranty benefits, that adapt to your research needs:
  • Interchange block types if your throughput needs change
  • Access the system remotely (and conveniently) through a mobile app
  • Program the instrument in seconds with a simple-to-use touchscreen interface
  • Simulate your old instrument with Thermal Simulation modes
  • Receive fast repair from factory-trained and certified service engineers
  • Minimize instrument downtime by troubleshooting with experienced Technical Support teams
For detailed instrument information and specifications, see the ProFlex PCR System (Cat. No. 4484075).
AB Repair Center (ABRC) Extended Warranty
Add additional years of warranty coverage to your existing manufacturer's limited warranty with the AB Repair Center (ABRC) extended warranty. With ABRC support, factory-trained engineers will maintain and repair your instrument at our remote service center. Just mail us your instrument and we'll maintain or repair it and ship it back to you within 3 weeks. All parts, labor, and shipping charges are included.
The ABRC extended warranty is ideal for customers in regulated environments that must maintain asset tag consistency to comply with regulated protocols.
Certain exclusions apply. Contact your Service and Support Representative for details at service.sales@thermofisher.com.
Specifications
For Use With (Equipment)ProFlex™ PCR System
Product LineProFlex
QuantitySystem + 1 yr REX ext warranty
Warranty Response TimeReplacement instrument shipped in 1 business day and received within 3 days
Warranty Services Not IncludedUser-replaceable Parts, Planned Maintenance
Warranty3 years of total service coverage with instrument rapid exchange
Unit SizeEach
Package Components
ProFlex™ 96-well PCR System
Catalog Number
4484075
Unit Size
1 instrument
Qty
1
ProFlex™ 96-Well PCR System, 1 year extended warranty, Rapid Exchange
Catalog Number
ZGEXESCPFLEX96W
Unit Size
1 yr ext warranty, Rapid Exchange
Qty
1