Search
Search
Bethyl Laboratories
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
SPAG5 (sperm associated antigen 5), also known as MAP126, hMAP126, DEEPEST or Astrin, is a 1,193 amino acid protein that is expressed at high levels in testis and at lower levels in liver, placenta, pancreas, colon and thymus. Localized specifically to spindle poles during prophase and throughout the spindle during metaphase and anaphase, SPAG5 is essential for proper spindle formation and is thought to be involved in the dynamic and functional regulation of mitotic spindles. Additionally, SPAG5 plays a role in sister chromatid cohesion and subsequent chromatid separation and, via its association with chromatids, is an important regulator of chromosome integrity. SPAG5 has a long stalk and a globular head domain through which if forms a homodimer that is characterized by aster-like structures. Upon DNA damage, SPAG5 may be phosphorylated by ATR or ATM.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Astrin; coiled-coil protein; Deepest; MAP126; mitotic spindle associated protein p126; mitotic spindle coiled-coil related protein; Mitotic spindle-associated protein p126; Sperm-associated antigen 5; unnamed protein product
Gene Aliases: DEEPEST; hMAP126; MAP126; SPAG5
UniProt ID: (Human) Q96R06
Entrez Gene ID: (Human) 10615
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support