Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • SUCLG2 Antibodies

          Bethyl Laboratories

          SUCLG2 Polyclonal Antibody

          View all (9) SUCLG2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SUCLG2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          SUCLG2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SUCLG2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SUCLG2 Antibody (A305-532A-T) in WB

          Detection of human and mouse SUCLG2 by western blot. Samples: Whole cell lysate (50 µg) from HeLa, HEK293T, Jurkat, mouse TCMK-1, and mouse NIH 3T3 cells prepared using NETN lysis buffer. Antibody: Affinity purified rabbit anti-SUCLG2 antibody (Product # A305-532A) (lot A305-532A-1) used for WB at 0.4 µg/mL. Detection: Chemiluminescence with an exposure time of 30 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SUCLG2 Antibody in Western Blot (WB)
          SUCLG2 Antibody in Immunoprecipitation (IP)
          SUCLG2 Polyclonal Antibody

          Product Details

          A305-532A-T

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:5,000
          -

          Immunoprecipitation (IP)

          2-10 µg/mg lysate
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Region between residue 25 to 75 of Human Succinyl-CoA ligase [GDP-forming] subunit beta, mitochondrial.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          10 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          tris citrate/phosphate, pH 7-8

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          The recommended shelf life for this product is 1 year from date of receipt.

          Based on 100% sequence identity, this antibody is predicted to react with Bovine and Pig

          Target Information

          This gene encodes a GTP-specific beta subunit of succinyl-CoA synthetase. Succinyl-CoA synthetase catalyzes the reversible reaction involving the formation of succinyl-CoA and succinate. Alternate splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 5 and 12.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: G-BETA; G-SCS; GTP-specific succinyl-CoA synthetase beta subunit; GTP-specific succinyl-CoA synthetase subunit beta; Itaconyl--CoA ligase [GDP-forming] subunit beta; Malyl--CoA ligase [GDP-forming] subunit beta; SCS-betaG; Succinate--CoA ligase [GDP-forming] subunit beta, mitochondrial; succinate-CoA ligase GDP-forming beta subunit; succinyl-CoA ligase [GDP-forming] subunit beta, mitochondrial; succinyl-CoA ligase, GDP-forming, beta chain, mitochondrial; Succinyl-CoA synthetase beta-G chain; succinyl-CoA synthetase, beta-G chain; unnamed protein product

          View more View less

          Gene Aliases: AF171077; AW556404; D6Wsu120e; G-SCS; GBETA; GTPSCS; SCS-betaG; SUCLG2

          View more View less

          UniProt ID: (Human) Q96I99, (Mouse) Q9Z2I8

          View more View less

          Entrez Gene ID: (Human) 8801, (Mouse) 20917

          View more View less

          Function(s)
          nucleotide binding magnesium ion binding catalytic activity succinate-CoA ligase (ADP-forming) activity succinate-CoA ligase (GDP-forming) activity protein binding ATP binding GTP binding ligase activity GDP binding macromolecular complex binding metal ion binding acyl-CoA ligase activity succinate-CoA ligase activity succinate-semialdehyde dehydrogenase (NAD+) activity 3-oxo-2-(2'-pentenyl)cyclopentane-1-octanoic acid CoA ligase activity fatty acid ligase activity 3-isopropenyl-6-oxoheptanoyl-CoA synthetase activity 2-oxo-delta3-4,5,5-trimethylcyclopentenylacetyl-CoA synthetase activity benzoyl acetate-CoA ligase activity 2,4-dichlorobenzoate-CoA ligase activity pivalate-CoA ligase activity cyclopropanecarboxylate-CoA ligase activity adipate-CoA ligase activity citronellyl-CoA ligase activity mentha-1,3-dione-CoA ligase activity thiophene-2-carboxylate-CoA ligase activity 2,4,4-trimethylpentanoate-CoA ligase activity cis-2-methyl-5-isopropylhexa-2,5-dienoate-CoA ligase activity trans-2-methyl-5-isopropylhexa-2,5-dienoate-CoA ligase activity branched-chain acyl-CoA synthetase (ADP-forming) activity aryl-CoA synthetase (ADP-forming) activity 3-hydroxypropionyl-CoA synthetase activity protein heterodimerization activity perillic acid:CoA ligase (ADP-forming) activity perillic acid:CoA ligase (AMP-forming) activity (3R)-3-isopropenyl-6-oxoheptanoate:CoA ligase (ADP-forming) activity (3R)-3-isopropenyl-6-oxoheptanoate:CoA ligase (AMP-forming) activity pristanate-CoA ligase activity malonyl-CoA synthetase activity ligase
          Process(es)
          tricarboxylic acid cycle succinyl-CoA metabolic process succinate metabolic process succinyl-CoA catabolic process metabolic process oxidation-reduction process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline