Alexa Fluor™ 647 Hydroxylamine
Invitrogen™

Alexa Fluor™ 647 Hydroxylamine

Alexa Fluor™ 647 Hydroxlamine is useful as a cell tracer and as a reactive dye for labeling aldehydes or ketonesRead more
Catalog NumberQuantity
A30632
also known as A-30632
1 mg
Catalog number A30632
also known as A-30632
Price (GBP)
241.00
Each
Quantity:
1 mg
Price (GBP)
241.00
Each
Alexa Fluor™ 647 Hydroxlamine is useful as a cell tracer and as a reactive dye for labeling aldehydes or ketones in polysaccharides or glycoproteins. Alexa Fluor™ 647 is a bright, far red fluorescent dye with excitation ideally suited to the 633 nm laser line. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 647 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 647 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

Detailed information about this AlexaFluor™ hydroxlamine:

• Fluorophore label : Alexa Fluor™ 647 dye
• Reactive group: hydroxlamine
• Reactivity: Aldehydes or ketones
• Ex/Em of the conjugate: 651/672 nm
• Extinction coefficient: 250,000 cm-1M-1
• Spectrally similar dyes: APC, Cy5
• Molecular weight: ∼1,200

Cell Tracking and Tracing Applications
Alexa Fluor™ hydrazides and hydroxlamines are useful as low molecular weight, membrane-impermeant, aldehyde-fixable cell tracers, exhibiting brighter fluorescence and greater photostability than cell tracers derived from other spectrally similar fluorophores. They are easily loaded into cells by microinjection, infusion from patch pipette, or uptake induced by our Influx™ Pinocytic Cell-Loading Reagent. Learn more about cell tracking and tracing.

Glycoprotein and Polysaccharide Labeling Applications
The Alexa Fluor™ hydrazides and hydroxlamines are reactive molecules that can be used to add a fluorescent label to biomolecules containing aldehydes or ketones. Aldehydes and ketones can be introduced into polysaccharides and glycoproteins by periodate-mediated oxidation of vicinal diols. Galactose oxidase can also be used to oxidize terminal galactose residues of glycoproteins to aldehydes.

Hydrazide vs Hydroxylamine
Hydrazine derivatives react with ketones and aldehydes to yield relatively stable hydrazones. Hydroxylamine derivatives (aminooxy compounds) react with aldehydes and ketones to yield oximes. Oximes are superior to hydrazones with respect to hydrolytic stability. Both hydrazones and oximes can be reduced with sodium borohydride (NaBH4) to further increase the stability of the linkage.

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

Related Products
DMSO (dimethylsulfoxide) (D12345)
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityCarboxylic Acid, Ketone, Aldehyde
Emission672 nm
Excitation651 nm
Label or DyeAlexa Fluor™ 647
Product TypeHydroxylamine
Quantity1 mg
Reactive MoietyAmine, Hydroxylamine
Shipping ConditionRoom Temperature
Label TypeAlexa Fluor
Product LineAlexa Fluor
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (4)

Citations & References
Abstract
New aldehyde tag sequences identified by screening formylglycine generating enzymes in vitro and in vivo.
Authors:Rush JS, Bertozzi CR,
Journal:J Am Chem Soc
PubMed ID:18722427
'Formylglycine generating enzyme (FGE) performs a critical posttranslational modification of type I sulfatases, converting cysteine within the motif CxPxR to the aldehyde-bearing residue formylglycine (FGly). This concise motif can be installed within heterologous proteins as a genetically encoded ' ... More
Site-specific chemical modification of recombinant proteins produced in mammalian cells by using the genetically encoded aldehyde tag.
Authors:Wu P, Shui W, Carlson BL, Hu N, Rabuka D, Lee J, Bertozzi CR,
Journal:Proc Natl Acad Sci U S A
PubMed ID:19202059
'The properties of therapeutic proteins can be enhanced by chemical modification. Methods for site-specific protein conjugation are critical to such efforts. Here, we demonstrate that recombinant proteins expressed in mammalian cells can be site-specifically modified by using a genetically encoded aldehyde tag. We introduced the peptide sequence recognized by the ... More
Enzyme-catalyzed transfer of a ketone group from an S-adenosylmethionine analogue: a tool for the functional analysis of methyltransferases.
Authors:Lee BW, Sun HG, Zang T, Kim BJ, Alfaro JF, Zhou ZS,
Journal:J Am Chem Soc
PubMed ID:20196537
'S-adenosylmethionine (AdoMet or SAM)-dependent methyltransferases belong to a large and diverse family of group-transfer enzymes that perform vital biological functions on a host of substrates. Despite the progress in genomics, structural proteomics, and computational biology, functional annotation of methyltransferases remains a challenge. Herein, we report the synthesis and activity of ... More
Introducing genetically encoded aldehydes into proteins.
Authors:Carrico IS, Carlson BL, Bertozzi CR
Journal:Nat Chem Biol
PubMed ID:17450134
Methods for introducing bioorthogonal functionalities into proteins have become central to protein engineering efforts. Here we describe a method for the site-specific introduction of aldehyde groups into recombinant proteins using the 6-amino-acid consensus sequence recognized by the formylglycine-generating enzyme. This genetically encoded 'aldehyde tag' is no larger than a His(6) ... More