Alexa Fluor™ 555 Cadaverine
Invitrogen™

Alexa Fluor™ 555 Cadaverine

Alexa Fluor™ 555 Cadaverine is useful as a polar tracer and as a reactive dye for labeling proteins via a자세히 알아보기
Have Questions?
카탈로그 번호수량
A306771 mg
카탈로그 번호 A30677
제품 가격(KRW)
507,000
온라인 행사
Ends: 30-Sep-2026
596,000
할인액 89,000 (15%)
Each
수량:
1 mg
제품 가격(KRW)
507,000
온라인 행사
Ends: 30-Sep-2026
596,000
할인액 89,000 (15%)
Each
Alexa Fluor™ 555 Cadaverine is useful as a polar tracer and as a reactive dye for labeling proteins via a carboxylic acid moiety. Alexa Fluor™ 555 is a bright, orange dye. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 555 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 555 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

Detailed information about this AlexaFluor™ cadaverine:

• Fluorophore label : Alexa Fluor™ 555 dye
• Reactive group: cadaverine
• Reactivity: carboxylic acids, aldehydes, and ketones (and glutamine residues through an enzyme-catalyzed transamidation reaction)
• Ex/Em of the conjugate: 555/572 nm
• Extinction coefficient: 155,000 cm-1M-1
• Spectrally similar dyes: TRITC
• Molecular weight: ∼950

Cell Tracking and Tracing Applications
Alexa Fluor™ cadaverines make excellent fluorescent polar tracers because they are bright, small, and water soluble. Since they contain an aldehyde-fixable functional group, they can be fixed in cells by treatment with formaldehyde or glutaraldehyde. They are easily loaded into cells by microinjection, infusion from patch pipette, or uptake induced by our Influx™ Pinocytic Cell-Loading Reagent. Learn more about cell tracking and tracing.

Protein Labeling Applications
Alexa Fluor™ cadaverines can be used as reactive molecules for adding a fluorescent label to carboxylic acids using a coupling agent such as a carbodiimide; they do not spontaneously react with carboxylic acids in solution. They do, however, react spontaneously with the common amine-reactive functional groups, including succinimidyl esters and isothiocyanates. The amine-containing Alexa Fluor™ cadaverines can also be used to label glutamine residues in some proteins and peptides via an enzyme-catalyzed transamidation reaction.

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

Related Products
DMSO (dimethylsulfoxide) (D12345)
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)
For Research Use Only. Not for use in diagnostic procedures.
사양
화학물질 반응성Carboxylic Acid, Ketone, Aldehyde
방출572 nm
여기555 nm
라벨 또는 염료Alexa Fluor™ 555
제품 유형Cadaverine
수량1 mg
반응성 부분Amine, Cadaverine
배송 조건Room Temperature
라벨 유형Alexa Fluor
제품라인Alexa Fluor
Unit SizeEach
구성 및 보관
Store in freezer -5°C to -30°C and protect from light.

인용 및 참조 문헌 (4)

인용 및 참조 문헌
Abstract
Visualization of galectin-3 oligomerization on the surface of neutrophils and endothelial cells using fluorescence resonance energy transfer.
Authors:Nieminen J, Kuno A, Hirabayashi J, Sato S
Journal:J Biol Chem
PubMed ID:17082191
'Galectin-3, a member of the galectin family of carbohydrate binding proteins, is widely expressed, particularly in cells involved in the immune response. Galectin-3 has also been indicated to play a role in various biological activities ranging from cell repression to cell activation and adhesion and has, thus, been recognized as ... More
SOI optical microring resonator with poly(ethylene glycol) polymer brush for label-free biosensor applications.
Authors:De Vos K, Girones J, Popelka S, Schacht E, Baets R, Bienstman P,
Journal:Biosens Bioelectron
PubMed ID:19200711
Label-free monitoring of biomolecular interactions has become of key importance for the emerging proteomics field. Monitoring real time interaction kinetics and high throughput screening of complex samples is of major importance for a variety of applications. We previously reported the use of Silicon-on-Insulator photonics microring resonators for cheap disposable biosensors ... More
Whole-cell 3D STORM reveals interactions between cellular structures with nanometer-scale resolution.
Authors:Huang B, Jones SA, Brandenburg B, Zhuang X,
Journal:Nat Methods
PubMed ID:19029906
The ability to directly visualize nanoscopic cellular structures and their spatial relationship in all three dimensions will greatly enhance our understanding of molecular processes in cells. Here we demonstrated multicolor three-dimensional (3D) stochastic optical reconstruction microscopy (STORM) as a tool to quantitatively probe cellular structures and their interactions. To facilitate ... More
In vivo imaging of membrane-associated glycans in developing zebrafish.
Authors:Laughlin ST, Baskin JM, Amacher SL, Bertozzi CR,
Journal:Science
PubMed ID:18451302
Glycans are attractive targets for molecular imaging but have been inaccessible because of their incompatibility with genetically encoded reporters. We demonstrated the noninvasive imaging of glycans in live developing zebrafish, using a chemical reporter strategy. Zebrafish embryos were treated with an unnatural sugar to metabolically label their cell-surface glycans with ... More