Minor Variant Finder Software (USB Format)
Applied Biosystems™

Minor Variant Finder Software (USB Format)

Minor Variant Finder Software is simple, easy-to-use desktop software designed for the accurate detection and reporting of minor variants inLeia mais
Have Questions?
Número do catálogoQuantity
A311001 each
Número do catálogo A31100
Preço (BRL)
-
Quantity:
1 each

Minor Variant Finder Software is simple, easy-to-use desktop software designed for the accurate detection and reporting of minor variants in Sanger sequencing traces with a detection level as low as 5%. Sophisticated algorithms filter out systemic noise components in bi-directional sequencing traces to quickly and accurately highlight and present genuine somatic minor variant candidates for review and reporting. This version of the software is provided in USB format. For digital download format, please see Cat. No. A30835.

Confidently call variants as low as 5%
The innovative algorithms of the Minor Variant Finder Software remove the background noise by comparing test sample(s) with a normal control sample. The algorithms produce a list of candidate variants and cleaned-up electropherograms for users to confidently review and confirm candidate variants on both forward and reverse strands. Review indicators facilitate user review of candidate minor variants detected during analysis.

The software can identify 5% minor variants with a sensitivity ≥ 95% and specificity ≥ 99% within the region of overlap among the forward and reverse test and control .ab1 files.

Simple and easy integration
The simple, workflow-based user interface guides users through each step from setup to review. Standard protocols for fluorescent dye terminator Sanger sequencing can be used, requiring little to no change. The minor variant candidates are presented for review in a convenient viewer tool and can be reported in a .csv and .vcf output file.

This desktop software supports .ab1 files generated from Applied Biosystems™ 3500, 3500xl, 3730, 3730xl, 3130, and 3130xl genetic analyzer instrument systems.

Complementary NGS confirmation
For high-throughput labs using nextgeneration sequencing (NGS) technology, minor variants from NGS findings can be confirmed with Sanger sequencing quickly and cost-effectively. The confirmation data can be visualized in an alignment view and Venn diagram for reporting.

Minor Variant Finder Software uses the GRCh38 human reference genome.

Computer requirements
Windows™ computer with 2 GB hard disk space and a minimum of 4 GB memory; 8 GB recommended.

  • Intel™ Core i7-4790 CPU, 3.60 Hz, Windows™ 10 Pro
  • Intel™ Core2 DUO CPU E8400, 3.00 GHz, Windows™ 7 SP1 32-bit
  • Intel™ Core i3-2100 CPU, 3.10 Hz (Windows™ 7 SP1 64-bit)
  • Intel™ Core i5-3380M, 2.9 GHz (Windows™ 7 SP1 64-bit)

Operating system:
Windows™ 7 SP1, 32-bit or 64-bit or Windows™ 10 Pro, 64-bit

Browser:
Google™ Chrome™, Mozilla™ Firefox™, Microsoft™ Internet Explorer™ v.11, or Microsoft™ Edge

The Minor Variant Finder Software runs in a web browser window, but does not require connection to the internet in order to run. Data is secure on your desktop computer.

Screen resolution:
1024x768 or higher, optimized for 1280x1024

For Research Use Only. Not for use in diagnostic procedures.

Especificações
For Use With (Equipment)3500 Genetic Analyzer
Includes1 USB media stick
Product TypeCE Secondary Analysis
Quantity1 each
FormatUSB
Unit SizeEach

Frequently asked questions (FAQs)

Can I use Minor Variant Finder Software to verify (Next-Generation Sequencing) NGS data?

Yes, Minor Variant Finder Software can be used to verify NGS data.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.

In what file formats can I export the variants using Minor Variant Finder Software?

Variants can be exported in .vcf and .csv file formats.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.

Does Minor Variant Finder Software allow easy review of detected variants?

Yes, the software allows reviewing of the original sequences of the test and control samples, as well as the noise-reduced sequence, all on the same page.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.

What sequencing files are needed to analyze a sample in Minor Variant Finder Software?

Forward and reverse sequences for the sample, as well as forward and reverse sequences for a control sample with the same primary sequence are needed to analyze a sample in Minor Variant Finder Software.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.

How does Minor Variant Finder Software work?

Minor Variant Finder Software subtracts sequencing background noise to allow for detection of rare minor variants.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.