Minor Variant Finder Software (USB Format) - FAQs
Minor Variant Finder Software (USB Format) - FAQs
View additional product information for Minor Variant Finder Software (USB Format) - FAQs (A31100)
7 product FAQs found
Can I use Minor Variant Finder Software to verify (Next-Generation Sequencing) NGS data?
Yes, Minor Variant Finder Software can be used to verify NGS data.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
In what file formats can I export the variants using Minor Variant Finder Software?
Variants can be exported in .vcf and .csv file formats.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Does Minor Variant Finder Software allow easy review of detected variants?
Yes, the software allows reviewing of the original sequences of the test and control samples, as well as the noise-reduced sequence, all on the same page.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What sequencing files are needed to analyze a sample in Minor Variant Finder Software?
Forward and reverse sequences for the sample, as well as forward and reverse sequences for a control sample with the same primary sequence are needed to analyze a sample in Minor Variant Finder Software.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How does Minor Variant Finder Software work?
Minor Variant Finder Software subtracts sequencing background noise to allow for detection of rare minor variants.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the sensitivity of the Minor Variant Finder Software?
Minor Variant Finder Software allows you to identify somatic mutations confidently down to 5% with 95% sensitivity and 99% specificity.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Do you have a video that shows how to use the Minor Variant Finder Software?
Yes, please see here (https://www.youtube.com/watch?v=hMbjhWzRQO8).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.