Is cell density in Floor Plate Specification medium critical for Dopaminergic neuron differentiation?
Yes, cell density should be optimized to attain full confluency at day 7 in the specification medium to have an efficient differentiation.