What substrate should I use with the PSC Definitive Endoderm Induction Kit?
We recommend using Vitronectin (VTN-N) Recombinant Human Protein, Truncated (Cat. No. A14700), but Geltrex LDEV-Free hESC-qualified Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1413301 or Cat. No. A1413302) can also be used.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
Can PSCs transitioned over into Essential 8 Medium on recombinant human laminin-521 (rhLaminin-521) from feeder-dependent cultures, then be transferred onto vitronectin (VTN-N)?
Yes. Following 2 passages on the rhLaminin-521 matrix, Versene or EDTA passaging should be used to subculture PSCs.
What are the main differences to be taken into consideration when culturing cells in Gibco Essential 8 Medium on Gibco Vitronectin (VTN-N) compared to other feeder-free systems?
Here are three major differences to be taken into consideration when culturing cells
in Gibco Essential 8 Medium on Gibco Vitronectin (VTN-N) compared to other feeder-free
systems:
- Cells should be typically passaged ~24 hours sooner than they would be with other feeder-free
media.
- Passaging should take place when cells are at ~85% confluency. If cells are passaged when they
are more than 85% confluent, the health of the cells and final cell yield may be compromised.
- Cells must be passaged in EDTA. Collagenase and dispase are not recommended.
Can pluripotent stem cells (PSCs) previously cultured in other media and on other substrates be cultured in Gibco Essential 8 Medium and on Gibco Vitronectin?
Cells cultured in other feeder-free media systems, such as mTeSR Medium with Matrigel Basement Membrane Matrix, or StemPro hESC SFM with Geltrex Matrix, can be
successfully cultured in Gibco Essential 8 Medium and VTN-N. In addition, PSCs grown on feeders with KnockOut SR have also been shown to be successfully cultured in Gibco Essential 8 Medium on VTN-N. However, when changing media systems, cells must be passaged either manually, or
with EDTA prior to culturing in Gibco Essential 8 Medium on VTN-N.
Can I use enzymes such as dispase and collagenase for passaging cells cultured in Gibco Essential 8 Medium and on VTN-N?
Enzymes such as dispase and collagenase do not work well with cells cultured in Gibco Essential 8 Medium on VTN-N. Use of these enzymes for passaging cells results in compromised viability and attachment.
Can pluripotent stem cells (PSCs) cryopreserved in a different culture condition be thawed and subsequently cultured in Gibco Essential 8 Medium and on vitronectin (VTN-N)?
Yes. PSCs cryopreserved from cultures of mTeSR Medium and BD Matrigel Basement Membrane Matrix may be thawed into Gibco Essential 8 Medium and plated on VTN-N. Certain lines may benefit from thawing into the medium and substrate they were growing in at the time of cryopreservation. Then at the next passage, use EDTA to passage the cells into Gibco Essential 8 Medium and VTN-N.
How should I thaw vitronectin (VTN-N)?
This product should be thawed at room temperature for 10-15 minutes. It should not be thawed at 4 degrees C overnight, which might result in poor product performance.
What temperature should be used to coat vitronectin (VTN-N) on plates?
We recommed coating the plate at room temperature for 1 hour.
What is the recommended passaging method to use with Gibco Essential 8 Medium and on Gibco Vitronectin (VTN-N)?
Cells cultured in Gibco Essential 8 Medium and VTN-N need to be passaged with EDTA.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
What will my cells look like growing in Gibco Essential 8 Medium and on Gibco Vitronectin (VTN-N)?
You should expect to see normal pluripotent stem cell (PSC) morphology. The expected morphology of PSCs is demonstrated specifically by tightly packed colonies with defined borders and a high nucleus-to-cytoplasm ratio.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
Can Gibco Essential 8 Medium and Gibco Vitronectin (VTN-N) support long-term growth of pluripotent stem cells (PSCs)?
Gibco Essential 8 Medium and vitronectin have been shown to support PSC growth for >50 passages without any signs of karyotypic abnormalities, and maintain the ability of PSCs to differentiate into all three germ line lineages. As published by Chen et al (http://www.ncbi.nlm.nih.gov/pubmed/21478862) in the laboratory of James Thomson, the VTN-N variant of vitronectin supports human pluripotent stem cell attachment and survival better than wild-type vitronectin when used in conjunction with Gibco Essential 8 Medium.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
Is Gibco Vitronectin (VTN-N) xeno-free (human or recombinant origin only)?
Yes. VTN-N is a defined, recombinant human protein.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
How does Gibco Vitronectin (VTN-N) compare to other basement membrane extracts?
Since VTN-N is a defined, recombinant human protein, variability is reduced in PSC cultures compared to human plasma-derived vitronectin and standard basement membrane extracts (BMEs). In addition, compared to full-length vitronectin and other defined substrates, VTN-N helps enable economical and scalable PSC culture.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.
What is Gibco Vitronectin (VTN-N)?
Gibco Vitronectin (VTN-N) is a recombinant, truncated human protein corresponding to the amino acid fragment 62-478 of human vitronectin expressed in E. coli. VTN-N is purified from inclusion bodies and refolded for use as a substrate for the feeder-free culture of human PSCs (http://www.ncbi.nlm.nih.gov/pubmed/21478862). When used with Gibco Essential 8 Medium, VTN-N has been proven to maintain pluripotency and normal growth characteristics in multiple PSC lines.
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.