KnockOut™ Serum Replacement - Multi-Species - FAQs
KnockOut™ Serum Replacement - Multi-Species - FAQs
View additional product information for KnockOut™ Serum Replacement - Multi-Species - FAQs (A3181502)
4 product FAQs found
How stable is the thawed KnockOut Serum Replacement Multi-Species when stored at 4 degrees C?
The product is stable for up to 4 weeks when stored at 4 degrees C and protected from light.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How many freeze/thaw cycles can KnockOut Serum Replacement Multi-Species go through?
The product can undergo one freeze/thaw cycle so we recommend thawing the vial and making single-use aliquots to refreeze.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
If I am currently using KnockOut Serum Replacement, can I switch to KnockOut Serum Replacement Multi-Species?
Yes. If you switch to KnockOut Serum Replacement Multi-Species for basic research applications, there will be little change to your experience with the product and it does not require regulatory documentation (or medical device clearance).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Is KnockOut Serum Replacement Multi-Species (Cat. Nos. A3181501 and A3181502) different from KnockOut Serum Replacement (Cat. Nos. 10828010 and 10828028)?
The formulation is the same for both products. KnockOut Serum Replacement Multi-Species is a Research Use Only product designed for use in basic research applications whereas KnockOut Serum Replacement is recommended for pre-clinical and clinical applications.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.