TaqMan™ Advanced miRNA Human A and B 96-well Plates, fast
Applied Biosystems™

TaqMan™ Advanced miRNA Human A and B 96-well Plates, fast

The Applied Biosystems TaqMan Advanced miRNA Human A and B 96-Well Plates, Fast, provide the convenience of pre-spotted TaqMan AdvancedRead more
Have Questions?
Catalog NumberQuantity
A318758 plates
Catalog number A31875
Price (USD)
2,585.00
Each
Quantity:
8 plates
Price (USD)
2,585.00
Each
The Applied Biosystems TaqMan Advanced miRNA Human A and B 96-Well Plates, Fast, provide the convenience of pre-spotted TaqMan Advanced miRNA assays in a 96-well fast plate format. This product contains a set of 8 plates for profiling of up to 752 unique miRNAs, and includes endogenous and exogenous miRNA controls for normalization of data results.

Features of the TaqMan Advanced miRNA Human A and B 96-Well Plates include:
• Reproducibility—pre-spotted assays are designed to reduce pipetting error and minimize experimental variability
• Sample savings—requires only 4 μL of purified plasma/serum or 2–20 ng of total RNA per sample
• Updated content—assay content updated for mirBase v.21
• Easy to use—just add a mix of diluted miR-Amp and master mix and run the qPCR; no additional pipetting required
• Gold-standard TaqMan qPCR chemistry—TaqMan Advanced miRNA assays draw on Thermo Fisher Scientific's industry-leading bioinformatics assay design pipeline and verification system to help ensure high specificity and minimal cross-reactivity, even for closely-related miRNA families

Product details:
Number of plates: 8
Number of unique miRNAs (not including exogenous controls): 752
Exogenous controls: cel-miR-39-3p, ath-miR159a
Endogenous control: hsa-miR-16-5p
Compatible with the following Applied Biosystems qPCR instruments: Viia7, StepOnePlus, 7500/7500 Fast, QuantStudio 3, QuantStudio 5, QuantStudio 6, QuantStudio 7 Flex, and QuantStudio 12k Flex
Recommended master mix: TaqMan Fast Advanced Master Mix

Notes
• TaqMan Advanced miRNA plates require the use of the TaqMan Advanced cDNA Synthesis Kit, which uses a universal reverse transcription (RT) chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA assays for detection and quantification of mature human miRNAs in biological samples.
• These plates are not compatible with the first-generation TaqMan MicroRNA cDNA Synthesis Kit, which uses an miRNA-specific reverse transcription chemistry.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7500 Fast System, 7900HT Fast System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System, StepOnePlus™ System
Format96-well plate
Product LineTaqMan
Product TypeAdvanced miRNA Human 96-Well Plate
Quantity8 plates
Sample TypeLiquid Samples (e.g. Serum), Tissue, Cells, FFPE & Fixed Samples, Blood
Shipping ConditionRoom Temperature
Volume4 μL
Detection MethodPrimer-probe
For Use With (Application)Microarray Analysis
GC-Rich PCR PerformanceHigh
PCR Method2-step RT-qPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

What is the minimum amount of TaqMan array plates I need to purchase to submit an order?

The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.

Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?

We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.

Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch

How do I obtain TaqMan documentation files for my TaqMan array plates?

Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number
• TaqMan Flexible Content panel array plates/Custom TaqMan array plates: Enter the Sales Order number and Batch number
• Click Submit.