TaqMan™ Advanced miRNA cDNA Synthesis Kit
Applied Biosystems™

TaqMan™ Advanced miRNA cDNA Synthesis Kit

Kit uses 3' poly-A tailing and 5' ligation of an adaptor sequence to extend the mature miRNAs present in the sample on each end prior to reverse transcription.
Have Questions?
카탈로그 번호수량
A2800750 cDNA synthesis reactions
카탈로그 번호 A28007
제품 가격(KRW)
762,000
온라인 행사
Ends: 31-Dec-2026
896,000
할인액 134,000
Each
수량:
50 cDNA synthesis reactions
제품 가격(KRW)
762,000
온라인 행사
Ends: 31-Dec-2026
896,000
할인액 134,000
Each

The Applied Biosystems TaqMan Advanced miRNA cDNA Synthesis Kit uses a universal reverse transcription (RT) chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA Assays for detection and quantification of mature miRNAs in biological samples. The kit is designed to perform 50 cDNA synthesis reactions, with theresulting cDNA sufficient for up to 600 qPCR reactions, allowing extensive downstream analysis.

Features of the TaqMan Advanced miRNA workflow include:

  • Sensitivity: high sensitivity allows for quantification of low-expressing miRNA targets, especially in biological fluids such as plasma, serum, and tissue
  • Flexibility: the universal RT chemistry generates universal cDNA templates for all miRNAs present in the sample, allowing for flexibility in study design
  • Sample savings: requires only 2 μL of plasma/serum sample input and generates enough cDNA for 600 qPCR reactions (20 μL) or 1200 qPCR reactions (10 μL)
  • Specificity: 5' ligation step allows for single base pair discrimination along entire miRNA sequence, including difficult-to-detect mismatches on 5' end
  • Gold-standard TaqMan chemistry: This kit is designed for use with TaqMan Advanced miRNA Assays for qPCR, which draw on Thermo Fisher Scientific's industry-leading bioinformatics assay design pipeline and validation system to help ensure high specificity and minimal cross-reactivity, even for closely-related miRNA families.

The TaqMan Advanced miRNA cDNA Synthesis Kit uses 3' poly-A tailing and 5' ligation of an adaptor sequence to extend the mature miRNAs present in the sample on each end prior to reverse transcription. Universal RT primers recognize the universal sequences present on both the 5' and 3' extended ends of the mature miRNAs. All mature miRNAs in the sample are reverse transcribed to cDNA.

In order to improve detection of low-expressing miRNA targets, the cDNA is then amplified using the Universal miR-Amp Primers and miR-Amp Master Mix to uniformly increase the amount of cDNA for each target, maintaining the relative differential expression levels. Unlike traditional pre-amplification, the Universal miR-Amp Primers recognize the universal sequences added to all mature miRNAs on the 5' and 3' ends, ensuring that there is no amplification bias.

Note: the TaqMan Advanced miRNA cDNA Synthesis Kit is not compatible with first generation TaqMan MicroRNA Assays for qPCR. For reverse transcription of miRNA targets using miRNA-specific RT primers, please see the TaqMan MicroRNA Reverse Transcription Kit.

For Research Use Only. Not for use in diagnostic procedures.
사양
용도(장비)7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System
반응 수50 cDNA synthesis reactions / 600 qPCR reactions
최적 반응 온도42°C
제품라인TaqMan
제품 유형cDNA Synthesis Kit
수량50 cDNA synthesis reactions
반응 형식Separate Components
역전사 효소SuperScript™ III
샘플 종류RNA
배송 조건Dry Ice
부피10 μL
검출 방법Primer-probe
용도(애플리케이션)miRNA analysis
GC-Rich PCR PerformanceHigh
PCR 방법qPCR
반응 속도Standard
Unit SizeEach
구성 및 보관
• 10X PolyA Buffer
• ATP, 10mM
• Poly A Enzyme, 5 U/μL
• 5X Ligase Buffer
• RNA Ligase, 10 U/μL
• 50% PEG 8000
• 25X Ligation Adaptor
• 10X RT Enzyme Mix
• 20X Universal RT Primer
• dNTP Mix, 100mM
• 20X miR-Amp Primer Mix
• 5X RT Buffer
• 2X miR-Amp Master Mix

Store all at -5 to -30°C, except 2X miR-Amp Master Mix which is stored at 2–8°C.

자주 묻는 질문(FAQ)

Can I purchase any of the components within the TaqMan Advanced miRNA cDNA Synthesis Kit (Cat. No. A28007) separately?

No, none of the single components within the kit are available as stand-alone. They only can be ordered within the complete kit.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

Do you have a modified protocol for the TaqMan MicroRNA Cells-to-CT Kit (Cat. No. 4391848) to make it compatible with the TaqMan Advanced miRNA Assays?

The workflow to use the Cell-to Ct kit with the TaqMan Advanced miRNA Assays is to use 2 µl of the lysate post cell lysis as input into the TaqMan Advanced miRNA cDNA synthesis Kit.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

What is the concentration of the probe and primer in the 20X TaqMan Advanced miRNA Assay (Cat. No. A25576)?

Unfortunately, these concentrations cannot be disclosed. Use the assay at a final concentration of 1x in the total reaction volume.

Find additional tips, troubleshooting help, and resources within our TaqMan Primers and Probes Support Center.

Do you have any suggestion on how to use the TaqMan Advanced miRNA Assay with Fluidigm system?

Our recommendation for this process would be to carry out a cDNA synthesis according to the TaqMan Advanced miRNA protocol, followed by scaling down QPCR reaction in accordance with the Fluidigm protocol.

At what concentration should I add the spike-in control into my sample for the TaqMan Advanced miRNA Assay?

The final concentration of the spike-in control in your test sample should be 1–10 pM.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.