SeqStudio™ Integrated Capillary Protector - FAQs
SeqStudio™ Integrated Capillary Protector - FAQs
View additional product information for SeqStudio™ Integrated Capillary Protector - FAQs (A31923)
2 product FAQs found
What is included in the SeqStudio Cartridge v2 (Cat. No. A41331) and can the cartridge (Cat. No. A41328) be purchased separately?
The SeqStudio Cartridge v2 includes the SeqStudio Genetic Analyzer Cartridge v2 and a SeqStudio Integrated Capillary Protector (Cat. No. A31923). Even though the cartridge has "A41328" on the box, it cannot be purchased separately and is only available as part of the SeqStudio Cartridge v2.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What does the SeqStudio Starter Kit (PN A35000) contain?
The SeqStudio Starter Kit (PN A35000) includes:
- SeqStudio Integrated Capillary Protector (Cat. No. A31923)
- SeqStudio Cathode Buffer Container (Cat. No. A33401)
- Sequencing Standard, BigDye Terminator v3.1, for 3500/SeqStudio Flex (Cat. No. 4404312)
- DS-33 GeneScan Installation Standards with GeneScan 600 LIZ Size Standard v2.0 (Cat. No. 4376911)
- Hi-Di Formamide (Cat. No. 4401457)
- MicroAmp Optical 96-Well Reaction Plate with Barcode (Cat. No. 4306737)
- MicroAmp 96-Well Tray/Retainer Set (Cat. No. 403081)
- MicroAmp Optical 8-Tube Strip, 0.2 mL (Cat. No. 4316567)
- Nuclease-Free Water (not DEPC-Treated) (Cat. No. AM9930)
- Cathode Buffer Container Reservoir Septa for SeqStudio Genetic Analyzer (Cat. No. A35640)
- 96-well Septa for SeqStudio Genetic Analyzer (Cat. No. A35641)
- 8-strip Septa for SeqStudio Genetic Analyzer (Cat. No. A35643)
- SeqStudio Cartridge v2 (PN A41328) (if this component is ordered online the catalog number is A41331)
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.