Product Image
Applied Biosystems™

SeqStudio™ Integrated Capillary Protector

The Applied Biosystems SeqStudio Integrated Capillary Protector (ICP) is used on the SeqStudio Cartridge. The Integrated Capillary Protector protects the자세히 알아보기
Have Questions?
카탈로그 번호수량
A319231 each
카탈로그 번호 A31923
제품 가격(KRW)
18,000
온라인 행사
Ends: 31-Dec-2026
19,000
할인액 1,000 (5%)
Each
수량:
1 each
제품 가격(KRW)
18,000
온라인 행사
Ends: 31-Dec-2026
19,000
할인액 1,000 (5%)
Each
The Applied Biosystems SeqStudio Integrated Capillary Protector (ICP) is used on the SeqStudio Cartridge. The Integrated Capillary Protector protects the capillary ends on the cartridge. The protector is gel-filled and is for one-time-use only. Store at 2–8°C before use.
For Research Use Only. Not for use in diagnostic procedures.
사양
수량1 each
배송 조건Approved for shipment at room temperature or wet ice
제품라인SeqStudio
유형Integrated Capillary Protector
Unit SizeEach
구성 및 보관
Store in refrigerator (2–8°C).

자주 묻는 질문(FAQ)

What is included in the SeqStudio Cartridge v2 (Cat. No. A41331) and can the cartridge (Cat. No. A41328) be purchased separately?

The SeqStudio Cartridge v2 includes the SeqStudio Genetic Analyzer Cartridge v2 and a SeqStudio Integrated Capillary Protector (Cat. No. A31923). Even though the cartridge has "A41328" on the box, it cannot be purchased separately and is only available as part of the SeqStudio Cartridge v2.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What does the SeqStudio Starter Kit (PN A35000) contain?

The SeqStudio Starter Kit (PN A35000) includes:
- SeqStudio Integrated Capillary Protector (Cat. No. A31923)
- SeqStudio Cathode Buffer Container (Cat. No. A33401)
- Sequencing Standard, BigDye Terminator v3.1, for 3500/SeqStudio Flex (Cat. No. 4404312)
- DS-33 GeneScan Installation Standards with GeneScan 600 LIZ Size Standard v2.0 (Cat. No. 4376911)
- Hi-Di Formamide (Cat. No. 4401457)
- MicroAmp Optical 96-Well Reaction Plate with Barcode (Cat. No. 4306737)
- MicroAmp 96-Well Tray/Retainer Set (Cat. No. 403081)
- MicroAmp Optical 8-Tube Strip, 0.2 mL (Cat. No. 4316567)
- Nuclease-Free Water (not DEPC-Treated) (Cat. No. AM9930)
- Cathode Buffer Container Reservoir Septa for SeqStudio Genetic Analyzer (Cat. No. A35640)
- 96-well Septa for SeqStudio Genetic Analyzer (Cat. No. A35641)
- 8-strip Septa for SeqStudio Genetic Analyzer (Cat. No. A35643)
- SeqStudio Cartridge v2 (PN A41328) (if this component is ordered online the catalog number is A41331)

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.