Alexa Fluor™ 488 Streptavidin, 5 nm colloidal gold conjugate - FAQs
Alexa Fluor™ 488 Streptavidin, 5 nm colloidal gold conjugate - FAQs
View additional product information for Alexa Fluor™ 488 Streptavidin, 5 nm colloidal gold conjugate - FAQs (A32360)
1 product FAQs found
I want to label F-actin with phalloidin and image with transmission electron microscopy. What is the best way to go about it?
One way is to use a biotin-phalloidin conjugate, then label that with a colloidal gold conjugate of streptavidin. You will need to block endogenous biotin first. Another method is to label with fluorescein phalloidin, then with an anti-fluorescein antibody conjugated with colloidal gold.