DataSafe™ Solution
Das tatsächliche Produkt kann abweichen
Ion Torrent™

DataSafe™ Solution

Have Questions?
Katalognummer A32634
Preis (EUR)
-
DataSafe™ Solution supports your laboratory’s big-data lifecycle. It includes storage consultation services, a network storage device with disk-failure tolerance, and ongoing storage administrator support. Key to this is the integration and configuration of your next-generation sequencing (NGS), molecular analysis, imaging, and other instrumentation with the DataSafe storage system, saving you setup time, costs, and burden on your internal IT team.

Benefits:

Turnkey
• Zero load on your team; rely on our storage consultants for storage hardware and white glove services
• Includes hardware integration, backup & replication services, definition of access rights, and other advanced configurations
• Includes one year of storage administrator support and hardware support

Protection
• Physical, on-site device with two disk-failure tolerance
• Controlled access to your laboratory results
• Automated backup and replication services

Scalability
• Private storage space for your laboratory and instrument data
• Compatible with NGS, real-time PCR, Sanger sequencing, imaging, and other laboratory data
• Scales as you grow—24 TB Solution can be expanded to 48 TB; 48 TB Solution can be expanded to 96 TB

Speed
• Transfer rates up to 100X faster than off-site or cloud storage
• Make your instrument run like new by freeing up on-instrument storage

What data types and instruments does this support?
DataSafe Solution allows you to share, store, and back up all of the data in your lab. For example, this cross-platform solution can be used to store NGS (Ion Torrent and 3rd party), qPCR, Sanger sequencing, and imaging data. Access is enabled from the PCs and laptops that you specify on your network.

Why do I need data storage and backup?
The loss of data from a single run can incur thousands of dollars of cost in lost reagents, time, and precious samples. External hard drives and USB flash drives are highly vulnerable to malfunction, damage, and theft. DataSafe Solutions minimizes the risk of losing valuable data with a scalable, networked storage solution.

Services and support
Service and support plans—comprehensive plans to maximize instrument performance and ensure availability of critical systems
Training—courses for experimental design, data analysis, troubleshooting, applications, and instrument operation
Bioinformatics—tailored support for NGS informatics execution, software use, data analysis, and downstream applications
DataSafe Solution—storage and back-up strategy, design, hardware, and support for all lab data

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Ion PGM™ System
Product LineDataSafe™
Product Size48 TB
Shipping ConditionRoom Temperature
TypeComputer
Unit Size48 TB
Inhalt und Lagerung
Turnkey storage and backup solution includes:
• 1 Dell® T630 network-attached storage appliance
• 48 TB usable RAID storage
• Integration services and support for NGS, qPCR, Sanger sequencing, imaging systems, and lab PCs
• Configuration services and support for backup, disaster recovery, advanced volume management, and export policies
• Training course on Ion Torrent data management
• Tower or rack mounting
• 1 year on-site hardware support
• 1 year of system adminstrator support

Häufig gestellte Fragen (FAQ)

How do I set up a network file sharing (NFS) to archive or backup data from Torrent Suite Software?

Settings depend on the file server that you have acquired. The destination folder must have read and write permissions. If you are using network file sharing (NFS), which we recommend, the “no_root_squash” option must be turned on.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How can I backup my data in Torrent Suite Software?

We provide a Torrent NAS (Network Attached Server - https://www.thermofisher.com/order/catalog/product/A32198. We can also provide other backup devices tailored to your needs - see more at https://www.thermofisher.com/order/catalog/product/A27662.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.