DataSafe™ Solution - FAQs
DataSafe™ Solution - FAQs
View additional product information for DataSafe™ Solution - FAQs (A32634)
2 product FAQs found
How do I set up a network file sharing (NFS) to archive or backup data from Torrent Suite Software?
Settings depend on the file server that you have acquired. The destination folder must have read and write permissions. If you are using network file sharing (NFS), which we recommend, the “no_root_squash” option must be turned on.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How can I backup my data in Torrent Suite Software?
We provide a Torrent NAS (Network Attached Server - https://www.thermofisher.com/order/catalog/product/A32198. We can also provide other backup devices tailored to your needs - see more at https://www.thermofisher.com/order/catalog/product/A27662.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.