Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 488 - FAQs
Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 488 - FAQs
View additional product information for Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 488 - FAQs (A32731, A32731TR)
3 product FAQs found
What mounting media should I use with samples labeled with Alexa Fluor Plus secondary antibodies?
All Alexa Fluor Plus secondary antibodies are compatible with the SlowFade Diamond and ProLong Diamond Antifade Mountants.
How do I use Alexa Fluor Plus secondary antibodies relative to the original Alexa Fluor secondary antibodies?
Because they are brighter, you would most likely be able to use a lower final concentration of the Alexa Fluor Plus secondary antibodies relative to the original Alexa Fluor secondary antibodies.
How are the Alexa Fluor Plus secondary antibodies different from the original Alexa Fluor secondary antibodies?
Compared to existing Alexa Fluor secondary antibodies, the new Alexa Fluor Plus secondary antibodies use a new proprietary dye chemistry coupled to highly cross-adsorbed antibodies, offering higher sensitivity and signal-to-noise ratios.