Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Secondary Antibodies ›
          • Goat anti-Rabbit IgG Secondary Antibodies
          Alexa Fluor Plus: high sensitivity, optimized for multiplexing and detection of low-abundance targets. Learn more

          Invitrogen

          Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 488

          44 Published Figures
          View all (241) Goat anti-Rabbit IgG antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Questions & Answers
          Tech Support

          Cite Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 488

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (21)
          • Published Figures (44)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 65

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody (A32731) in ICC/IF

          Immunofluorescent analysis of MAP2 in the differentiated neurons from H9 ESC-derived NSCs. 2 weeks after differentiation, cells were fixed, permeabilized and stained with a MAP2 rabbit polyclonal antibody (Product # PA5-17646) at 1:100 dilution (green) and a HuC/HuD mouse monoclonal antibody (Product # A-21271), at a concentration of 5 µg/mL (red) in blocking buffer for at least 1 hour at room temperature, ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Western Blot (WB)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (IHC)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Western Blot (WB)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Western Blot (WB)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Western Blot (WB)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Western Blot (WB)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          A32731

          Applications
          Tested Dilution

          Western Blot (WB)

          0.1-0.4 µg/mL

          Immunohistochemistry (Paraffin) (IHC (P))

          1:2,000

          Immunocytochemistry (ICC/IF)

          1-10 µg/mL
          Product Specifications

          Species Reactivity

          Rabbit

          Host/Isotype

          Goat / IgG

          Class

          Polyclonal

          Type

          Secondary Antibody

          Immunogen

          Gamma Immunoglobins Heavy and Light chains

          Conjugate

          Alexa Fluor™ Plus 488 Alexa Fluor™ Plus 488 Alexa Fluor™ Plus 488
          • Alexa Fluor 350 
          • Alexa Fluor 405 
          • Alexa Fluor 430 
          • Alexa Fluor 488 
          • Alexa Fluor 488-10 nm colloidal gold
          • Alexa Fluor 488-5 nm colloidal gold
          • Alexa Fluor 514 
          • Alexa Fluor 532 
          • Alexa Fluor 546 
          • Alexa Fluor 555 
          • Alexa Fluor 568 
          • Alexa Fluor 594 
          • Alexa Fluor 633 
          • Alexa Fluor 635 
          • Alexa Fluor 647 
          • Alexa Fluor 660 
          • Alexa Fluor 680 
          • Alexa Fluor 680 
          • Alexa Fluor 700 
          • Alexa Fluor 750 
          • Alexa Fluor 790 
          • Alexa Fluor Plus 405 
          • Alexa Fluor Plus 555 
          • Alexa Fluor Plus 594 
          • Alexa Fluor Plus 647 
          • Alexa Fluor Plus 680 
          • Alexa Fluor Plus 750
          • Alexa Fluor Plus 800 
          • AP
          • AP
          • APC 
          • Biotin 
          • Biotin 
          • Biotin 
          • Biotin-XX
          • Cyanine3
          • Cyanine5 
          • DyLight 350
          • DyLight 405
          • DyLight 488
          • DyLight 550
          • DyLight 594
          • DyLight 633
          • DyLight 650
          • DyLight 680
          • DyLight 755
          • DyLight 800
          • DyLight 800 4X PEG
          • FITC
          • FITC
          • HRP
          • HRP
          • Oregon Green 488
          • Pacific Blue
          • Pacific Orange
          • PE 
          • PE-Alexa Fluor 610
          • PE-Alexa Fluor 647
          • PE-Cyanine5.5
          • Rhodamine
          • Request custom conjugation

          Excitation/Emission Max

          493/518 nm View spectra spectra

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          1 mg

          Purification

          Affinity chromatography

          Storage buffer

          proprietary buffer, pH 6.5

          Contains

          0.016% Methylisothiazolone, 0.016% Bromonitrodioxane

          Storage conditions

          4°C, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2633280

          Target

          IgG

          Cross Adsorption

          Against bovine IgG, goat IgG, mouse IgG, rat IgG and human IgG

          Antibody Form

          Whole Antibody

          Product Specific Information

          To minimize cross-reactivity, the goat anti-rabbit IgG whole antibodies have been pre cross-adsorbed against bovine IgG, goat IgG, mouse IgG, rat IgG, and human IgG. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in less background staining and cross-reactivity. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. Further passages through additional columns result in highly cross-adsorbed preparations of secondary antibody. The benefits of these extra steps are apparent in multiplexing/multicolor-staining experiments where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins.

          Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically.

          Target Information

          Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          View product citations for antibody A32731 on CiteAb

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-xbl4w:80/100.66.75.228:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0