MicroSEQ&trade; ID <i>Ultra</i> Sequencing Clean-up 8-Strips Refill Kit
Applied Biosystems™

MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Refill Kit

The MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit is used for post-sequencing clean-up in the MicroSEQ ID workflow.
Have Questions?
Catalog NumberQuantity
A3325696 Samples
Catalog number A33256
Price (BRL)
-
Quantity:
96 Samples

The MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Refill Kit is used for post-sequencing clean-up in the MicroSEQ™ ID workflow. It includes the 8-well strips that are part of the MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246) but does not include the 96-well receiver plate. The gel filtration matrix used in the strip cartridges provides a convenient and simple way to remove dye terminators, dNTPs, salts, and residual primers from cycle sequencing reactions.

The MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit can be used downstream of the following sequencing kits:

  • MicroSEQ™ 500 16S rDNA Sequencing Kit
  • MicroSEQ™ Full Gene 16S rDNA Sequencing Kit
  • MicroSEQ™ D2 rDNA Fungal Sequencing Kit

The kit consists of twelve 8-well strips suitable for 96 sequencing samples. It is ideal for low- to medium-throughput microbial-identification workflows. The end products can be used directly for sequencing on any Applied Biosystems™ genetic analyzers.

Specifications
For Use With (Application)Sequencing
High-throughput CompatibilityMultiplexing
Product LineMicroSEQ™ ID
Purification TargetFluorescent Dye Removal
Quantity96 Samples
FormatStrips
Sample TypeSequencing Products
Unit SizeEach
Contents & Storage
Sufficient for 96 sequencing samples

12 x 8-strips clean-up tubes

Frequently asked questions (FAQs)

How do Phasemaker Tubes work?

Phasemaker Tubes work on the principle of density gradient centrifugation. Phasemaker Tubes contain a thick liquid polymer that is heavier than the aqueous phase of the TRIzol Reagent mix, but lighter than the organic phase, so after centrifugation, the polymer positions itself between these two layers and forms an exclusive physical barrier. It separates and completely isolates the upper aqueous phase from the organic phase underneath, making it very easy to aspirate the upper phase containing the RNA.

How long is the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256) stable for?

The product is guaranteed stable for one year when stored properly at 4 degrees C.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

Can I store the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256) at room temperature (20-24 degrees C)?

Yes. The kit contents are stable when stored in their original packaging at room temperature for up to 4 weeks. However, the recommended storage condition is at 4 degrees C.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

How many sequencing samples is the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256) sufficient for?

The MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit is sufficient for 96 sequencing samples.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

What are the catalog numbers for the MicroSEQ ID Ultra Sequencing Clean-up Kits?

Here the kits with the catalog numbers:

- MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246)
- MicroSEQ ID Ultra Sequencing Clean-up Plates Kit (Cat. No.A33245)
- MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256)

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.