MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Refill Kit - FAQs
MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Refill Kit - FAQs
View additional product information for MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Refill Kit - FAQs (A33256)
9 product FAQs found
How do Phasemaker Tubes work?
Phasemaker Tubes work on the principle of density gradient centrifugation. Phasemaker Tubes contain a thick liquid polymer that is heavier than the aqueous phase of the TRIzol Reagent mix, but lighter than the organic phase, so after centrifugation, the polymer positions itself between these two layers and forms an exclusive physical barrier. It separates and completely isolates the upper aqueous phase from the organic phase underneath, making it very easy to aspirate the upper phase containing the RNA.
How long is the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256) stable for?
The product is guaranteed stable for one year when stored properly at 4 degrees C.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
Can I store the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256) at room temperature (20-24 degrees C)?
Yes. The kit contents are stable when stored in their original packaging at room temperature for up to 4 weeks. However, the recommended storage condition is at 4 degrees C.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
How many sequencing samples is the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256) sufficient for?
The MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit is sufficient for 96 sequencing samples.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
What are the catalog numbers for the MicroSEQ ID Ultra Sequencing Clean-up Kits?
Here the kits with the catalog numbers:
- MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246)
- MicroSEQ ID Ultra Sequencing Clean-up Plates Kit (Cat. No.A33245)
- MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256)
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
What is included in the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256)?
The kits consists of twelve 8-well strips suitable for 96 sequencing samples.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
How long is the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips good for use once opened?
Strips should be used within 2 weeks of opening. We recommend returning unused strips from the 96-well plate holder to the foil package, sealing, and storing at 4 degrees C.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
What is the minimum reaction volume for use of the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256)?
The minimum reaction volume is 5 µL. The reaction volume should be brought up to at least 5 µL with distilled water before adding to the 8-well strips.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
What is the storage condition for the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256)?
The kit is stable when stored for 12 months at 4 degrees C or for 4 weeks at ambient temperature (20-24 degrees C).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.