PI(5)P Lipid Kinase Substrate
Invitrogen™
PI(5)P Lipid Kinase Substrate
The PI(5)P Lipid Kinase Substrate has been specifically optimized for use with Type II PIP kinases (e.g., PIP4K2A). The substrateRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A33593400 nmol
A335965000 nmol
Catalog number A33593
Price (MXN)
-
Quantity:
400 nmol
The PI(5)P Lipid Kinase Substrate has been specifically optimized for use with Type II PIP kinases (e.g., PIP4K2A). The substrate consists of solubilized phosphatidylinositol 5-phosphate (PI(5)P) in an aqueous buffer, and is used in combination with our Adapta™ Universal Kinase Assay to help identify unique kinase inhibitors.
• Validated for use with Thermo Fisher Scientific lipid kinases
• Available in multiple pack sizes for convenient use
• Also available in our SelectScreen™ Kinase Profiling Service
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineInvitrogen
Quantity400 nmol
Shipping ConditionDry Ice
SubstratePhosphatidylinositol 5-phosphate (PI(5)P)
FormPowder
Substrate TypeKinase Substrate
Unit SizeEach
Contents & Storage
Phosphatidylinositol 5-phosphate (PI(5)P) in 30 mM HEPES pH 7.5 buffer.