PI(3,4)P2 Lipid Kinase Substrate
Invitrogen™
PI(3,4)P2 Lipid Kinase Substrate
The PI(3,4)P2 Lipid Kinase Substrate has been specifically optimized for use with Type I PIP kinases (e.g., PIP5K1A, PIP5K1B, PIP5K1C).Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A33597400 nmol
A335985000 nmol
Catalog number A33597
Price (USD)
1,232.00
Each
Quantity:
400 nmol
Price (USD)
1,232.00
Each
The PI(3,4)P2 Lipid Kinase Substrate has been specifically optimized for use with Type I PIP kinases (e.g., PIP5K1A, PIP5K1B, PIP5K1C). The substrate consists of solubilized phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) in an aqueous buffer, and is used in combination with our Adapta™ Universal Kinase Assay to help identify unique kinase inhibitors.
• Validated for use with Thermo Fisher Scientific lipid kinases
• Available in multiple pack sizes for convenient use
• Also available in our SelectScreen™ Kinase Profiling Service
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineInvitrogen
Quantity400 nmol
Shipping ConditionDry Ice
SubstratePhosphatidylinositol 3,4-bisphosphate (PI(3,4)P2)
FormPowder
Substrate TypeKinase Substrate
Unit SizeEach
Contents & Storage
Phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) in 30 mM HEPES pH 7.5 buffer.