The PI(3,4)P2 Lipid Kinase Substrate has been specifically optimized for use with Type I PIP kinases (e.g., PIP5K1A, PIP5K1B, PIP5K1C).Read more
Have Questions?
Change view
Catalog Number
Quantity
A33598
5000 nmol
A33597
400 nmol
2 Options
Catalog number A33598
Price (KRW)
10,659,000
Each
Quantity:
5000 nmol
Price (KRW)
10,659,000
Each
The PI(3,4)P2 Lipid Kinase Substrate has been specifically optimized for use with Type I PIP kinases (e.g., PIP5K1A, PIP5K1B, PIP5K1C). The substrate consists of solubilized phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) in an aqueous buffer, and is used in combination with our Adapta™ Universal Kinase Assay to help identify unique kinase inhibitors.