RPMI 1640 Medium for SILAC
Thermo Scientific™

RPMI 1640 Medium for SILAC

Thermo Scientific™ RPMI 1640 Medium for SILAC is optimized for use with stable isotope labeling with amino acids in cell자세히 알아보기
Have Questions?
카탈로그 번호수량
A338236 x 500 mL
카탈로그 번호 A33823
제품 가격(KRW)
388,000
온라인 행사
Ends: 30-Sep-2026
456,000
할인액 68,000 (15%)
Each
수량:
6 x 500 mL
제품 가격(KRW)
388,000
온라인 행사
Ends: 30-Sep-2026
456,000
할인액 68,000 (15%)
Each
Thermo Scientific™ RPMI 1640 Medium for SILAC is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).

Features of RPMI 1640 Medium for SILAC:
• Flexible—liquid media deficient in both L-lysine and L-arginine, allowing for more complete proteome coverage through multiple isotopic amino acid labeling
• High quality—medium is sterile, endotoxin-free, and cell culture-compatible

SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of 'heavy' 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization, and quantitation of proteins.

This SILAC medium is used together with heavy and light amino acids for the differential isotopic labeling of cells for SILAC analysis. SILAC media supplemented with heavy amino acids are capable of nearly 100% label incorporation into proteins of living cells.

Related products
NeuCode™ Lysine -080, 50 mg
Fetal Bovine Serum, dialyzed, US origin
DMEM for SILAC

For Research Use Only. Not for use in diagnostic procedures.
사양
최종 제품 유형Liquid Medium
라벨 또는 염료Mass Isotope of Amino Acid
물질Cell Lysate
수량6 x 500 mL
배송 조건Room Temperature
형태Liquid
제품 유형Media
시작 물질Cell Lysate
Unit SizeEach
구성 및 보관
Store at 2–8°C.