QuantStudio™ 5 Real-Time PCR System for Human Identification, 96-well, 0.2 mL, laptop
Applied Biosystems™
QuantStudio™ 5 Real-Time PCR System for Human Identification, 96-well, 0.2 mL, laptop
The Applied Biosystems QuantStudio 5 Real-Time PCR System for Human Identification with laptop computer is the latest addition to ourRead more
Have Questions?
Catalog NumberQuantity
A343211 system
Catalog number A34321
Price (USD)
-
Quantity:
1 system
The Applied Biosystems QuantStudio 5 Real-Time PCR System for Human Identification with laptop computer is the latest addition to our human identification (HID) real-time PCR family of products. This instrument is designed for both new and experienced users who need a simple, reliable, and affordable real-time PCR system that doesn’t require compromise on performance or quality. When used with our latest advances in quantification chemistry and HID Real-Time PCR Analysis Software, this system offers a sensitive, robust solution for forensic DNA quantification. It also helps prepare laboratories for future quantification technologies. With maximum dye versatility, this HID solution offers accurate, trusted results in a small benchtop footprint.
Features of the QuantStudio 5 Real-Time PCR System for Human Identification:
Streamlined
• Smaller foot print: 27 W x 50 D x 40 H cm, designed to maximize horizontal and vertical bench space
• Front-facing fully motorized block
Interactive
• Touch screen interface enables viewing of amplification plots
• Easy and intuitive interface
Reliable
• Bright light LED median lifetime greater than 5 years
• Less frequent maintenance
• 10 GB on-board memory for storage of 2,000–5,000 runs
• HID-validated system
• 0.2 mL standard 96-well format
Innovative
• Multiplexing capability for up to six dyes, enabling future quantification technology
• Precise temperature control with Peltier-controlled blocks (known as VeriFlex zones)
• USB interface
Real-Time PCR applications
Real-time PCR is a flexible technique that can be used for multiple forensic applications:
• Forensic database
    --Laboratories without direct amplification workflows
• Forensic casework
    --Confirmatory screening of male DNA
    --Sexual assault sample workflow
    --Quality and quantity sample assessment
Specifications
Display TypeTouchscreen
Quantity1 system
Shipping ConditionRoom Temperature
Block FormatNon-interchangeable
Unit SizeEach
Contents & Storage
Laptop
Frequently asked questions (FAQs)
I am unable to download the current software update for the QuantStudio 5 Real-Time PCR System from the Thermo Fisher website. Why?
We recommend trying different browsers (Google Chrome, Firefox, and Microsoft Edge) and/or removing cookies from your browser. If this does not resolve the issue, please contact Technical Support at techsupport@thermofisher.com.
What is the admin pin for QuantStudio 3 and 5 Real-Time PCR Systems?
If the admin PIN has not been changed, 1234 should work. If it does not work, you can also try 0000.
However, if the admin PIN has been changed and none of the above PINs work, follow the instructions in the QuantStudio 3 and 5 Real-Time PCR Systems manual, page 100.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Is the black frame removable from the QuantStudio 3 and 5 Real-Time PCR Systems 96 well, 0.2 ml block?
We do not recommend physically removing the black frame. It is necessary for the plates to fit properly and should stay on the instrument. If it has been accidently removed or lost, please contact ICS to replace the spare part at (800) 9550-6288 options 3-2 or email technicalhardwaresupport@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
On which real-time PCR instruments can I use the Quantifiler Duo DNA Quantification Kit?
The Quantifiler Duo DNA Quantification Kit has been optimized and validated for use on the Applied Biosystems 7500 Real-Time PCR System and the Applied Biosystems Human Identification (HID) QuantStudio 5 Real-Time PCR System.
How does the HID Real-Time PCR Software v1.4 differ from the previous version, v1.3?
HID Real-Time PCR Software v1.4 includes the same feature set as version 1.3 and runs on Windows 10 as well. The difference is that HID Real-Time PCR software v1.4 is designed to be compatible with the latest QuantStudio 5 firmware version 1.5.1.