SeqStudio™ Genetic Analyzer System with SmartStart
Applied Biosystems™

SeqStudio™ Genetic Analyzer System with SmartStart

The Applied Biosystems SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis system designed to provide users with an integratedRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberContent And Storage
A35645
A35644
A35646
Catalog number A35645
Price (USD)
-
The Applied Biosystems SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis system designed to provide users with an integrated experience that is easy, efficient, and versatile for research in genetic analysis. The all-in-one reagent cartridge allows easy operation for multiple users with the flexibility to perform both sequencing and fragment analysis in a single run. The simplified SeqStudio Plate Manger software is well-suited for first-time and experienced users. Be more connected to your data than ever with intuitive browser-based software and cloud-based connectivity options for remote monitoring, and data transfer and analysis.

The SeqStudio system gives you the same data quality, reliability, and support you have come to expect from the Applied Biosystems brand with a modernized experience. Same workflow, same trusted data. Now with one-click convenience.

Key features
• Simplified instrument software with interactive touch screen for greater ease of use
• Minimized hands-on setup time with integrated reagents cartridge (SeqStudio Cartridge)
• Application flexibility to run Sanger sequencing and fragment analysis on the same run
• Connectivity: Wi-Fi or wired Ethernet connection
• Web-enabled remote monitoring capability
• Consumables usage tracking through radio frequency identification (RFID)

This package includes:
• 4-capillary SeqStudio Genetic Analyzer
• SeqStudio Analysis Software (GeneMapper, Sequencing Analysis, Minor Variant Finder, Variant Reporter, and SeqScape)
• SeqStudio Plate Manager Software
• 1Day SmartStart Orientation
Note: SeqStudio Cartridge and other system consumables and reagents sold separately

Just click. That's it.
Load up your samples, click in the cartridge, and go—no need for technical experts for setup or maintenance, no calibration before each run, and no installation of capillaries, polymer, and anode buffer. The cartridge is stable for 4 months on the instrument.

Access your data anytime, anywhere*
This system features Thermo Fisher Cloud connectivity so you can set up your run, check its progress, and access your data from wherever you have Internet access, all without the need to stay in the lab.* Get the same data quality, service, and support you’ve come to expect from Applied Biosystems technology with a modernized experience at a price any lab can afford.
* Internet connection and Thermo Fisher Cloud account required.

Service, support, and training
We offer a range of service options, plans, and warranties to improve productivity, increase instrument uptime, and lower the total cost of ownership. SmartStart orientation helps get you started with your new instrument more quickly.

Software
SeqStudio Genetic Analyzer Instrument Software, v 1.1Software update fileRelease notesEULA
SeqStudio Genetic Analyzer Plate Manager Software, v1.1Windows installerMac installer
Sequence Analysis Software patch for SeqStudio supportInstallerRelease notes 
SeqScape Software patch for SeqStudio supportInstallerRelease notes 
Variant Reporter Software patch for SeqStudio supportInstallerRelease notes 
GeneMapper Client Software patch for SeqStudio supportInstallerRelease notes 
GeneMapper Full Software patch for SeqStudio supportInstallerRelease notes 
Minor Variant Finder Software patch for SeqStudio supportInstaller 


For Research Use Only. Not for use in diagnostic procedures.
Specifications
Duration1 Year Extended Warranty
Shipping ConditionRoom Temperature
Product LineSeqStudio
TypeGenetic Analyzer
Unit SizeEach
Package Components
A34274
Catalog Number
A34274
Unit Size
Qty
1
4443764
Catalog Number
4443764
Unit Size
Qty
1
A34684
Catalog Number
A34684
Unit Size
Qty
1
ZG61SCSEQSTUDIO
Catalog Number
ZG61SCSEQSTUDIO
Unit Size
Qty
1
Contents & Storage
System + SmartStart + 1 yr ext warranty

Frequently asked questions (FAQs)

Have you tested the SeqStudio Genetic Analyzer (Cat. No. A35644, A35645) in low humidity conditions (below 20%)?

SeqStudio Genetic Analyzer has not been tested for operation in low humidity conditions and is validated for environments with 20-80% humidity. Therefore, we do not recommend using SeqStudio Genetic Analyzer in below 20% humidity.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

If the computer attached to the SeqStudio Genetic Analyzer System with SmartStart is upgraded to Win 10, does the data collection software need to be upgraded as well?

No, since the SeqStudio instrument's data collection software is on the instrument itself, it is not necessary that you upgrade it.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.

Can I run SNaPshot applications, between 35 and 120 bp, on the SeqStudio Genetic Analyzer and the Applied Biosystems 3500 Genetic Analyzer with a 50 cm capillary array?

Both the SeqStudio Genetic Analyzer and the Applied Biosystems 3500 Genetic Analyzer have run modules specific for the SNaPshot application.

- For the 3500 Genetic Analyzer, the run module is "SNaPshot50_POP7" and it has a ~30 minute run time.
- For the SeqStudio Genetic Analyzer, the run module is "SNaPshot", the optimal size range is 40-120 bp, and it runs for ~25 minutes but the 35 bp fragment will be collected.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.

How do I set up the same user, system security, audit, and e-signature settings on multiple instances of the SAE Administrator Console v2.0?

You may use the export and import functions to transfer settings from one installation of the SAE Administrator Console to another.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.

What does archiving audit records in the SAE Administrator Console do?

Archiving audit records removes the records from the SAE Administrator Console and saves them in an internally specified location on the same computer on which the SAE Administrator Console is installed. Note: Archived audit records are accessible for viewing in the SAE Administrator Console.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.