What were the results of comparability studies between the original ExoSAP-IT Reagent and the improved ExoSAP-IT Express Reagent provided with the MicroSEQ ID Purification Combo Kit v2.0?
Samples treated with the ExoSAP-IT and ExoSAP-IT Express reagents produced very similar sequencing results and the correct identification results.
Is ExoSAP-IT Express Reagent available as a stand-alone product?
Yes. The unit sizes available range from 480 to 2,000 reactions (https://www.thermofisher.com/order/catalog/product/75001.1.EA).
What is the minimum reaction volume to be used with the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit, Cat. No. A33246?
The minimum reaction volume is 5 µL. The reaction volume should be brought to at least 5 µL with distilled water before adding to the 8-well strips.
How long are the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips good for once opened?
Strips should be used within 2 weeks of opening. We recommend storing unused strips from the 96-well plate holder, in the foil package, sealing and storing at 4 degrees C.
What are the storage conditions for the MicroSeq ID Purification Combo Kit v2.0, with clean-up plates?
Here are the storage conditions:
-ExoSAP-IT Express Reagent: Store at -5 to -30 degrees C
-MicroSEQ ID Ultra Sequencing Clean-up Plate: Store at 2-8 degrees C in a non-frost free freezer
How is the MicroSEQ ID Purification Combo Kit v2.0 shipped?
It is shipped on wet or dry ice.
What is the catalog number for the MicroSeq ID Purification Combo Kit v2.0, with clean-up plates?
It is Cat. No. A35852.
What is included with the MicroSEQ ID Purification Combo Kit v2.0, with clean-up plates?
Here are the components:
-ExoSAP-IT Express Reagent (one tube, sufficient for 100 reactions)
-MicroSEQ ID Ultra Sequencing Clean-up Plate (one 96-well plate, sufficient for 96 reactions)
Will primer-dimers affect the efficiency of ExoSAP-IT Express Reagent used in the MicroSEQ ID Purification Combo Kit v2.0?
Yes, ExoSAP-IT Express Reagent cannot digest primer-dimers because they are double-stranded.
What is the difference between ExoSAP-IT Reagent vs ExoSAP-IT Express Reagent included in the MicroSEQ ID Purification Combo Kit v2.0?
The difference is in the incubation time. ExoSAP-IT Reagent has a 15 min incubation step at 37 degrees C and another 15 min incubation at 80 degrees C. ExoSAP-IT Express Reagent has a total 5 min protocol with a 4 min incubation step at 37 degrees C followed by a 1 min incubation at 80 degrees C.