LIF Recombinant Mouse Protein, embryonic stem cell-qualified, 100 μg - FAQs
LIF Recombinant Mouse Protein, embryonic stem cell-qualified, 100 μg - FAQs
View additional product information for LIF Recombinant Mouse Protein, embryonic stem cell-qualified - FAQs (A35933, A35934, A35935)
2 product FAQs found
What are the differences between Leukemia Inhibitory Factor (LIF) Recombinant Mouse Protein, Embyonic Stem Cell-Qualified and LIF Recombinant Human Protein?
Besides the species difference between LIF Recombinant Mouse Protein, Embyonic Stem Cell-Qualified (Cat. Nos. A35933, A35934, A35935) and LIF Recombinant Human Protein (Cat. No. PHC9484), the other main difference is that ESC-qualified mouse LIF is derived from plant tissue, whereas LIF Recombinant Human Protein is produced in E. coli. ESC-qualified mouse LIF is verified to contain less than 0.005 ng/µg of endotoxin, which is more than ten times lower than that in E. coli-derived recombinant human LIF protein. These two recombinant proteins also have different reconstitution instructions, different storage conditions, and shelf life.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can Leukemia Inhibitory Factor (LIF) Recombinant Mouse Protein, Embyonic Stem Cell-Qualified work with human cells?
No, Leukemia Inhibitory Factor (LIF) Recombinant Mouse Protein, Embryonic Stem Cell-Qualified cannot work in human cells as it does not bind human LIF receptor.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.