LIF Recombinant Mouse Protein, embryonic stem cell-qualified
Gibco™

LIF Recombinant Mouse Protein, embryonic stem cell-qualified

Gibco embryonic stem cell (ESC)–qualified mouse LIF is a recombinant protein that supports the culture and maintenance of mouse pluripotentRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A35935100 μg
A3593450 μg
A3593310 μg
Catalog number A35935
Price (USD)
1,728.00
Each
Quantity:
100 μg
Price (USD)
1,728.00
Each

Gibco embryonic stem cell (ESC)–qualified mouse LIF is a recombinant protein that supports the culture and maintenance of mouse pluripotent stem cells (PSCs) by suppressing spontaneous differentiation. Produced in barley grain to minimize endotoxin levels, each lot is carefully tested for purity, microbial contamination, bioactivity, and ability to support mouse PSC culture, thereby minimizing the risk of contamination and helping ensure robust performance.

With LIF Recombinant Mouse Protein, ESC-Qualified, you can:
• Rely on a recombinant growth factor that is qualified for mouse PSC culture
• Safeguard mouse PSC quality with a low-endotoxin LIF
• Culture with confidence with a rigorously tested reagent

Rely on an ESC-qualified growth factor
LIF bioactivity is typically measured by observing its ability to induce differentiation of murine M1 myeloid leukemia cells. In addition to this standard assay, each lot of ESC-qualified mouse LIF is tested for its ability to maintain Oct4 expression in mouse PSCs, thus helping to ensure robust performance for mouse PSC culture.

Safeguard mouse PSC quality with a low-endotoxin LIF
Unlike bacteria-derived recombinant proteins which may carry over harmful bacterial endotoxins, ESC-qualified mouse LIF is derived from plant tissue. It is verified to contain less than 0.005 ng/μg of endotoxin, more than ten times lower than most bacteria-derived counterparts. This unique, low-endotoxin LIF supports the optimal conditions for safeguarding the health and quality of your mouse PSC culture.

Culture with confidence with a rigorously tested reagent
ESC-qualified mouse LIF is tested for bacteria and mycoplasma to minimize the risk of contamination. It is analyzed by SDS-PAGE to ensure >95% purity. It is also tested for pyrogenic and pro-inflammatory contaminants via monocyte activation test.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ClassificationES Cell-Qualified
Expression SystemHordeum vulgare (Barley Grain)
Gene SymbolLIF
Protein FamilyCytokines & Receptors
Protein FormRecombinant
Protein TagHis Tag
Purity or Quality Grade>95% by SDS-PAGE
Quantity100 μg
Research CategoryStem Cell Research
Shipping ConditionRoom Temperature
For Use With (Application)Cell Culture
FormLyophilized
RecombinantRecombinant
SpeciesMouse
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).

Frequently asked questions (FAQs)

What are the differences between Leukemia Inhibitory Factor (LIF) Recombinant Mouse Protein, Embyonic Stem Cell-Qualified and LIF Recombinant Human Protein?

Besides the species difference between LIF Recombinant Mouse Protein, Embyonic Stem Cell-Qualified (Cat. Nos. A35933, A35934, A35935) and LIF Recombinant Human Protein (Cat. No. PHC9484), the other main difference is that ESC-qualified mouse LIF is derived from plant tissue, whereas LIF Recombinant Human Protein is produced in E. coli. ESC-qualified mouse LIF is verified to contain less than 0.005 ng/µg of endotoxin, which is more than ten times lower than that in E. coli-derived recombinant human LIF protein. These two recombinant proteins also have different reconstitution instructions, different storage conditions, and shelf life.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can Leukemia Inhibitory Factor (LIF) Recombinant Mouse Protein, Embyonic Stem Cell-Qualified work with human cells?

No, Leukemia Inhibitory Factor (LIF) Recombinant Mouse Protein, Embryonic Stem Cell-Qualified cannot work in human cells as it does not bind human LIF receptor.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.