High Select™ HSA/Immunoglobulin Depletion Mini Spin Columns, 6 Columns - FAQs
High Select™ HSA/Immunoglobulin Depletion Mini Spin Columns, 6 Columns - FAQs
View additional product information for High Select™ Depletion Spin Columns - FAQs (A36369, A36368, A36367, A36372, A36371, A36370, A36366, A36365)
3 product FAQs found
Can I use the High Select HSA/Immunoglobulin Depletion Resin to process mouse serum samples?
No, the High Select HSA/Immunoglobulin Depletion Resin cannot be used for mouse samples; the depletion resin contains immobilized antibodies that only recognize human serum or plasma proteins.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can I reuse the High Select HSA/Immunoglobulin Depletion Resin?
We don’t recommend reusing the High Select HSA/Immunoglobulin Depletion Resin as depletion efficiency may decrease after regeneration, and subsequent depleted samples may be contaminated from prior ones.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What formats of the High Select HSA/Immunoglobulin Depletion Resin do you offer?
The High Select HSA/Immunoglobulin Depletion Resin is offered in 3 formats:
a) Resin alone: High Select HSA/Immunoglobulin Depletion Resin (Cat. No. A36368)
b) High Select HSA/Immunoglobulin Depletion Mini Spin Columns (Cat. Nos. A36365, A36366) that can process a maximum of 10 µL of human serum or plasma
c) High Select HSA/Immunoglobulin Depletion Midi Spin Columns (Cat. No. A36367) that can process a maximum of 100 µL of human serum or plasma. For serum samples containing abnormally high amounts of albumin or IgG, the sample load may need to be reduced.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.