Oncomine™ Childhood Cancer Research Assay - FAQs
Oncomine™ Childhood Cancer Research Assay - FAQs
View additional product information for Oncomine™ Childhood Cancer Research Assay - FAQs (A36485)
7 product FAQs found
How many primer pools are in the Oncomine Childhood Cancer Research Assay panel?
There are 4 primer pools in total, two DNA and two RNA pools across 202 genes. This assay allows concurrent analysis of DNA and RNA to simultaneously detect multiple types of variants, including hotspots, single nucleotide variants (SNVs), indels, copy number variants (CNVs), and gene fusions, in a single workflow.
What is included in the Oncomine Childhood Cancer Research Assay panel?
The panel comprises 203 unique genes: 130 key DNA genes, 28 copy number variant targets, an expansive fusion panel of 90 driver genes with multiple partners, and 9 expression genes and controls. The panel also includes the Ion AmpliSeq Sample ID Panel primers to prevent research sample misidentification and provide gender assignment of the research sample. Please see this flyer: https://assets.thermofisher.com/TFS-Assets/CSD/Flyers/oncomine-childhood-cancer-assay-flyer.pdf
What controls can I run with the Oncomine Childhood Cancer Research Assay (Cat. Nos. A36485, A36486)?
The recommended controls are:
-Acrometrix Oncology Hotspot Control, Cat. No. 969056 (https://www.thermofisher.com/order/catalog/product/969056)
-Seraseq FFPE Tumor Fusion RNA Reference Material v2, Cat. No. 0710-0129 (https://www.seracare.com/Seraseq-FFPE-Tumor-Fusion-RNA-Reference-Material-v2-0710-0129/)
What is the starting material for the Oncomine Childhood Cancer Research Assay (Cat. No. A36485)?
This panel is compatible with as little as 10 ng per pool input DNA or RNA, per library, from blood, bone marrow, fresh/frozen tissue or FFPE samples. Reverse transcription of each sample requires 12 ng of DNase-treated total RNA (≥2.5 ng/µL) for manual library preparation and 2 × 5 ng/primer pool plus overage for the RT reaction. Each manual DNA library target amplification reaction requires 10 ng (≥1.82 ng/µL) of mammalian gDNA from normal or FFPE tissue per pool (20 ng in total).
With the Oncomine Childhood Cancer Research Assay, how many samples can be multiplexed on an Ion 540 chip?
We have validated 8 samples on an Ion 540 chip: 7 DNA samples plus one negative control PLUS 7 RNA samples plus one negative control per chip; RNA and DNA from the same sample; 16 barcodes on each chip (8 barcodes for DNA and 8 barcodes for RNA).
What is included in the Oncomine Childhood Cancer Research Assay (Cat. No. A36485)?
The Oncomine Childhood Cancer Research Assay (Cat. No. A36485) consists of the DNA Oncomine Childhood Cancer Research Panel (2-pools), RNA Oncomine Childhood Cancer Research Panel (2-pools), and two Ion AmpliSeq Library Kit Plus (Cat. No. 4488990) kits for the rapid preparation of barcoded sample libraries from DNA and RNA. Sufficient reagents are provided to prepare uniquely barcoded DNA- and RNA-libraries from 24 research samples.
What is the Oncomine Childhood Cancer Research Assay (Cat. Nos. A36485, A36486)?
The Oncomine Childhood Cancer Research Assay is a unique next-generation sequencing (NGS)-based assay designed for comprehensive genomic profiling of cancers affecting children and young adults. It consists of four pools of Ion AmpliSeq oligonucleotide primers and associated reagents to generate amplicon libraries for next-generation sequencing on the Ion GeneStudio S5 platforms. Since the driving genetic aberrations of these malignancies are different than those affecting adults, a specific and comprehensive panel is needed to advance research. The Oncomine Childhood Cancer Research Assay (Cat. No. A36485) is for manual library preparation where barcodes are not included, and the Oncomine Childhood Cancer Research Assay, Chef-Ready (Cat. No. A36486) is for automated library preparation using the Ion Chef System.