TrueCut™ Cas9 Protein v2, 100 μg - FAQs
TrueCut™ Cas9 Protein v2, 100 μg - FAQs
View additional product information for TrueCut™ Cas9 Protein v2 - FAQs (A36499, A36498, A36497, A36496)
7 product FAQs found
What is the molecular weight of TrueCut Cas9 Protein v2?
The molecular weight of TrueCut Cas9 Protein v2 is ~163 kD
What is the difference between TrueCut Cas9 Protein (Prototype) and TrueCutCas9 Protein v2?
Both products are the same CRISPR-Cas9 protein, but the TrueCut Cas9 Protein is manufactured with a new production process and formulated to meet the standards for Ancillary Materials for Cell and Tissue-Based Products.
Is there a specific antibody that you recommend to test the delivery of TrueCut Cas9 Protein v2?
Although we do not have a specific antibody recommendation to test the delivery of TrueCut Cas9 Protein v2, we have tested the following antibodies against an older version of Cas9 (Platinum Cas9) using U2OS-Cas9 expression cell lines (generated using Cas9 lentivirus):
- Cas9 Recombinant Polyclonal Antibody (Cat. No. 711061)
- Cas9 Monoclonal Antibody (Cat.No. MA1-201)
- Cas9 Monoclonal Antibody (Cat. No. MA1-202)
Our TrueCut Cas9 Protein v2 and the Platinum Cas9 have similar domains, and the TrueCut Cas9 Protein v2 has better efficiencies. Therefore, these antibodies should work with TrueCut Cas9 Protein v2 as well.
What is the storage buffer composition for TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein?
The storage buffer composition for TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein is as follows:
15 mM Tris-HCl pH 8.0, 150 mM NaCl, 1.0 mM TCEP, 50% glycerol.
Which lipid transfection reagent do you recommend using with TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein?
We recommend using Lipofectamine CRISPRMAX Transfection Reagent with TrueCut Cas9 Protein v2 or TrueCut HiFi Cas9 Protein.
Do you offer the TrueCut Cas9 Protein v2 at different concentrations?
Yes, we offer TrueCut Cas9 Protein v2 in the following concentrations: 1 µg/µL (Cat. Nos. A36496, A36497) for use in standard editing scenarios, 5 µg/µL (Cat. Nos. A36498, A36499) for optimization of editing conditions in more difficult scenarios such as in primary or embryonic cells, microinjection, or when screening multiple gRNA sequences at a time
What gRNA format do you recommend using with TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein?
TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein are designed to work with TrueGuide synthetic gRNAs, but can also be used with in vitro transcribed gRNA from the GeneArt Precision gRNA Synthesis Kit.