Ion AmpliSeq™ HD Dual Barcode Kit 1-24 - FAQs
Ion AmpliSeq™ HD Dual Barcode Kit 1-24 - FAQs
View additional product information for Ion AmpliSeq™ HD Dual Barcode Kit 1-24 - FAQs (A37695)
6 product FAQs found
Can the 96-well plate in the Ion AmpliSeq HD Dual Barcode Kit 1-24 (Cat. No. A37695) be freeze/thawed multiple times?
We have taken the 96-well plate in the Ion AmpliSeq HD Dual Barcode Kit 1-24 through several freeze-thaw cycles with little to no reduction in performance.
Note: Sealing the plate between uses is key when using the plate for multiple experiments.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What version of Torrent Suite Software do I need in order to run the Ion AmpliSeq HD Dual Barcode Kit 1-24?
The Ion AmpliSeq HD Dual Barcode Kit 1-24 is compatible with Torrent Suite Software version 5.10 and above.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Is the Ion AmpliSeq HD Dual Barcode Kit 1-24 shipped on ice or at room temperature?
The kit is shipped at room temperature, but should be stored at -20 degrees C.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many samples is one Ion AmpliSeq HD Dual Barcode Kit 1-24 good for?
The Ion AmpliSeq HD Dual Barcode Kit 1-24 is good for 24 samples. Each barcode is good for one sample.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Is the Ion AmpliSeq HD Dual Barcode Kit 1-24 compatible with the Ion Chef System?
No, it is only compatible with manual sample preparation.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the Ion AmpliSeq HD Dual Barcode Kit 1-24?
The Ion AmpliSeq HD Dual Barcode Kit 1-24 provides a set of 24 unique dual-barcode primers designed and validated for optimal performance with Ion GeneStudio S5 series sequencers. The kit enables up to 24 samples to be multiplexed on a single chip and improves the accuracy of multiplex sequencing by reducing sample mis-assignment. The barcode kit is only compatible with the Ion AmpliSeq HD Library Kit (Cat. No. A37694) and should not be used with any other library kit.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.