MiniAmp™ Thermal Cycler, 1 instrument - FAQs
MiniAmp™ Thermal Cycler, 1 instrument - FAQs
View additional product information for MiniAmp™ Thermal Cycler - FAQs (A38081, A38080, A37834)
9 product FAQs found
Why are my PCR tubes crushed/deformed after my PCR run?
The heated thermal cycler lid is designed to make sure that optimized pressure is applied on the microplate for an efficient PCR reaction. When using PCR tubes, excess pressure on the tubes, from the heated lid can cause deformation of the tubes. To avoid this, we recommend using the Tray/Retainer set, as shown below:
Cat. No. 4381850, MicroAmp 96-Well Tray/Retainer Set for Veriti Systems – This Tray is for use with MicroAmp Strip Tubes (or MicroAmp Reaction Tubes without Caps, 0.2 mL), with the Veriti Thermal Cycler, SimpliAmp Thermal Cycler, and Proflex PCR System.
Cat. No. 403081, MicroAmp 96-Well Tray/Retainer Set – This Tray is for use with MicroAmp Strip Tubes (or MicroAmp Reaction Tubes without Caps, 0.2 mL), with the 7000 System, 2720 Thermal Cycler, and GeneAmp PCR System 9700.
Cat. No. 4379983, MicroAmp 96-Well Tray for VeriFlex Blocks – This Tray is for use with MicroAmp Strip Tubes (or MicroAmp Reaction Tubes with Caps, 0.1 mL), with the Veriti Thermal Cycler, Veriti Fast Thermal Cycler, SimpliAmp Thermal Cycler, and Proflex PCR System.
Please take a look at this video: How to Use Adapter Tray Retainers in Thermal Cycler Starter Kits (https://www.youtube.com/watch?v=1uh2ZxZJtgs).
Find additional tips, troubleshooting help, and resources within our PCR Thermal Cyclers Support Center.
What is the optimal ramp rate for my thermal cycler?
There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.
As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.
Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf
Find additional tips, troubleshooting help, and resources within ourPCR Thermal Cyclers Support Center.
What is the difference between the MiniAmp and MiniAmp Plus Thermal Cyclers?
There are 2 key differences:
- The MiniAmp Plus Thermal Cycler has 3 VeriFlex zones while the MiniAmp Thermal Cycler does not.
- The MiniAmp Plus Thermal Cycler has a block ramp rate of 3.5 degrees C/sec while the MiniAmp Thermal Cycler has a block ramp rate of 3.0 degrees C/sec.
Is there wireless connectivity with the MiniAmp/MiniAmpPlus Thermal Cycler?
Like the ProFlex PCR System and SimpliAmp Thermal Cycler, the MiniAmp/MiniAmpPlus Thermal Cycler can be connected wirelessly with an optional High-Power USB W-Fi Module, Cat. No. A26774.
How do I program an infinite hold in the MiniAmp/MiniAmpPlus Thermal Cycler?
To add an infinite hold, select the infinity symbol for program.
What is the Auto Delta feature on the MiniAmp/MiniAmpPlus Thermal Cycler?
The AutoDelta feature allows you to incrementally increase or decrease the temperature or hold time with each cycle for a given step within a cycling stage. One common application for this is touchdown PCR.
What is the maximum block ramp rate of the MiniAmp/MiniAmp Plus Thermal Cycler?
The maximum block ramp rate of the MiniAmp Thermal Cycler is 3 degrees C/sec.
The maximum block ramp rate of the MiniAmp Plus Thermal Cycler is 3.5 degrees C/sec.
What types of plastics can I use with the MiniAmp/MiniAmp Plus Thermal Cycler?
The compatible plates and plastics are the same as those used with the ProFlex 96-Well PCR System and can be viewed here (https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/thermal-cycler-plastics/proflex-96-3x32.html).
What types of blocks are available on the MiniAmp/MiniAmp Plus Thermal Cycler?
We offer only a 96-well standard (0.2 mL) block that is not interchangeable. The 96-well block for the MiniAmp Plus Thermal Cycler has a block with 3 VeriFlex zones while the MiniAmp Thermal Cycler has an isothermal block.