Can the Ion GeneStudio S5 System be used for diagnostic procedures?
The Ion GeneStudio S5 System (Cat. No. A38194) is labeled For Research Use Only and is not intended for use in diagnostic procedures. Diagnostic workflows require instruments, assays, and software appropriate for the intended diagnostic use. Contact Technical Support for product-specific guidance.
How many reads can I expect from an Ion 540 Chip on the Ion S5 System?
An Ion 540 Chip on the Ion S5 System typically generates approximately 60-80 million reads per run. Actual output can vary by library type, run quality, and application. For pricing or run-cost estimates, contact your sales representative or refer to current quote documentation.
What software is needed to operate and monitor runs on the Ion GeneStudio S5 System?
The Ion GeneStudio S5 System uses the Torrent Suite Software to plan, monitor, track, and analyze sequencing runs. The Torrent Suite Software is pre-installed on the system.
What Ion Sphere Particle (ISP) format is used with Ion Chef workflows?
The Ion Sphere Particles (ISPs) used depend on the chip type selected for the workflow. Each Ion Chef kit includes ISPs that are specific to the corresponding chip type to support template preparation.
What is the difference between the Ion S5 System and the Ion GeneStudio S5 System?
The Ion S5 System and the Ion GeneStudio S5 System have the same functionality and support the same consumables, chip types, and assays. The Ion S5 System was rebranded as the Ion GeneStudio S5 System, and only the GeneStudio S5 System is currently manufactured.
Can I run the Ion GeneStudio S5 System/Ion Chef Instrument at lower humidity levels (10-20% humidity) rather than the 40% humidity recommended in the user guide?
We do not recommend running the Ion GeneStudio S5 System/Ion Chef Instrument in environmental conditions other than our recommended operating conditions. When the instrument runs within a lower humidity range of 10-20% there are risks to the electronics, as low humidity levels are likely to lead to electrostatic discharges. Damage to the electronics can result in run failure and hence performance cannot be guaranteed. Please ensure that the instrument is run at the recommended humidity of 40%.
Find additional tips, troubleshooting help, and resources within our
Next-Generation Sequencing Support CenterWith Ion S5 2X Cleaning Solution, how long does a "pulse clean" take to complete?
It takes approximately 47 mins to do a "pulse clean",
When using Ion S5 2X Cleaning Solution, where in the software should I select "pulse clean"?
Go to "System tools" and select "pulse clean". The instrument must already have been cleaned after a run.
What are the dimensions of the bottles in Ion S5 2X Cleaning Solution?
Here are the bottle dimensions:
- Length (Metric) 61 mm
- Height (Metric) 131.1 mm
- Width (Metric) 61 mm
How many bottles of Ion S5 2X Cleaning Solution do you provide in Cat. No. A34172?
The kit contains 2 bottles of Ion S5 2X Cleaning Solution.
Which instruments is Ion S5 2X Cleaning Solution compatible with?
Ion S5 2X Cleaning Solution is compatible with GeneStudio S5/GeneStudio S5 Plus/GeneStudio S5 Prime systems and Ion S5/Ion S5 XL systems.
When should I use Ion S5 2X Cleaning Solution?
Ion S5 2X Cleaning Solution is used to perform a pulse clean of your Ion S5 Sequencer (GeneStudio S5/GeneStudio S5 Plus/GeneStudio S5 Prime systems and Ion S5/Ion S5 XL systems) to either recover the instrument performance from a diagnosed solid-state buffer leak or perform preventative maintenance if the instrument has not been used for several months.
Is the Ion GeneStudio S5 System compatible with the Ion One Touch 2 System?
Is the Ion GeneStudio S5 System compatible with the Ion Chef System?
What is the server storage size on the Ion GeneStudio S5 System?
How long does analysis take on the Ion GeneStudio S5 System?
Please see the following table (https://www.thermofisher.com/us/en/home/life-science/sequencing/next-generation-sequencing/ion-torrent-next-generation-sequencing-workflow/ion-torrent-next-generation-sequencing-run-sequence/ion-s5-ngs-targeted-sequencing/ion-s5-specifications.html).
Find additional tips, troubleshooting help, and resources within our
Next-Generation Sequencing Support Center.
Can the Ion GeneStudio S5 System run two chips back-to-back?
No, the analysis of the first chip must complete before the sequencing run of the second chip begins.
Find additional tips, troubleshooting help, and resources within our
Next-Generation Sequencing Support Center.
Does the Ion GeneStudio S5 System have a separate analysis server?
How long does it take to run an Ion 540 Chip on the Ion GeneStudio S5 System?
What is the maximum throughput/day for the Ion GeneStudio S5 System?
What chips are compatible with the Ion GeneStudio S5 System?
Ion 510 Chip, Ion 520 Chip, Ion 530 Chip, and Ion 540 Chip are compatible with the Ion GeneStudio S5 System.
Find additional tips, troubleshooting help, and resources within our
Next-Generation Sequencing Support Center.