Ion GeneStudio™ S5 System
Ion Torrent™
Ion GeneStudio™ S5 System
The Ion GeneStudio™ S5 System is a semiconductor-based next-generation sequencing (NGS) system that enables simple, flexible targeted sequencing workflows for clinical research and research laboratories.
Have Questions?
Número do catálogoIncludes
A38194System only
Número do catálogo A38194
Preço (BRL)
-
Includes:
System only
The Ion GeneStudio™ S5 System is a semiconductor-based next-generation sequencing (NGS) system that enables simple, flexible targeted sequencing workflows for clinical research and research laboratories. From targeted gene panels to exomes and transcriptomes, the Ion GeneStudio S5 system helps laboratories manage both small and medium-throughput sequencing projects across oncology, inherited disease, reproductive health, infectious disease, microbial genomics, and transcriptomics applications.
With cartridge-based reagents and the flexible Ion 4-Series chip portfolio, the system is simple to use while providing scalable sequencing throughput. The Ion GeneStudio S5 system supports a broad range of targeted DNA sequencing and RNA sequencing applications, enabling laboratories to increase sample throughput or sequencing depth using the same proven workflow.
The Ion GeneStudio™ S5 system is the base model within the GeneStudio family that also includes the GeneStudio S5 and the Ion GeneStudio S5 Prime systems. It supports the 510 up to the 540 Chips, but it cannot run the largest 550 and 560 Chips, compatible only with the GeneStudio Plus or Prime.
Key features:
  • Select the chip (Ion 510 - 540 Chip) that fits your application and throughput needs to achieve from 2–80 million reads per run
  • Reduce setup time and complexity with cartridge-based plug-and-play reagents
  • Complete sequencing and primary analysis in hours to move experiments forward faster
The Ion GeneStudio™ S5 Series
The Ion GeneStudio™ S5 systems share the same foundational semiconductor sequencing technology and fluidic architecture across all three instruments: the Ion GeneStudio™ S5 system, the Ion GeneStudio™ S5 Plus system, and the Ion GeneStudio™ S5 Prime system. The primary difference across the three instruments is on-sequencer analysis speed and supported chips. While the GeneStudio™ S5 Plus and Prime systems can run any of the Chips, from the 510 to the 560, the GeneStudio™ base model can only support the 510, 520, 530 and 540 Chips.
Speed
The Ion GeneStudio S5 Series leverages the speed of semiconductor sequencing to produce high-quality sequencing data in just a few hours. When paired with the Ion Chef™ System, laboratories can go from library preparation to sequencing data in as little as 24 hours with approximately 45 minutes of total hands-on time.
Simplicity
With single-day installation, a straightforward user interface, and cartridge-based reagents, the Ion GeneStudio S5 systems are easy to use benchtop sequencers. Combined with Ion AmpliSeq™ technology for targeted panel design, the Ion Chef™ System for automated library and template preparation, and Ion Reporter™ Software for automated variant annotation, targeted NGS workflows are simpler than ever.
Small sample input
Low-input DNA and RNA from challenging sample types—including formalin-fixed paraffin-embedded (FFPE) tissue, fine needle aspirates, and circulating cell-free DNA and RNA (cfDNA/cfRNA)—can be difficult to sequence on other next-generation sequencing platforms. With Ion AmpliSeq technology and the Ion GeneStudio S5 Series, laboratories can sequence with as little as 1 ng of DNA or RNA input.
Scalability
Why purchase multiple sequencing systems when one platform can support all your targeted sequencing needs? The Ion 6 Series chip portfolio—including the new Ion 560 Chip—provides multiple throughput options so laboratories can efficiently scale from small research studies to high-volume targeted sequencing projects. Choose the sequencing output that best matches your application, from small targeted panels to comprehensive genomic profiling (CGP), transcriptomics, and custom AmpliSeq™ workflows, while maintaining the same simple workflow.
Simple analysis solutions
Torrent Suite™ Software and Ion Reporter™ Software make NGS data analysis accessible for both new and experienced users. Plan, monitor, track, and analyze sequencing runs using Torrent Suite Software (pre-installed on the Sequencer). Integrate, annotate, and interpret variants using Ion Reporter Software, available as either a cloud-based or local deployment. LIMS connectivity services, including API training and implementation support, help laboratories integrate sequencing workflows into existing laboratory information systems.
Services and support
With more than 2,000 field personnel worldwide, Thermo Fisher Scientific provides one of the industry's largest global networks of Field Application Specialists and Service Engineers to support your laboratory throughout implementation and routine operation.
  • Service plans:—Instrument service agreements designed to support routine operation and system availability.
  • Qualification services:—Installation and operational qualification services that document instrument performance according to manufacturer specifications.
  • Training:—Instructor-led and virtual courses covering instrument operation, workflow setup, troubleshooting, and NGS applications.
  • Bioinformatics support:—Resources for workflow implementation, software configuration, data analysis, and downstream applications.
For Research Use Only. Not for use in diagnostic procedures.
Especificações
Analysis Time1.5 to 19 hr. BAM File Generation (Depending on the read length and Chip size)
Automated WorkflowModular, Combined with the Ion Chef for library prep and Chip loading
Capacity∼12 TB
Compatibility510, 520, 530 and 540 Chips
Dimensions (L x W x H) Exterior80.6 x 54.2 x 50.9 cm
For Use With (Application)Targeted Oncology, Reproductive Health, Microbiology, Inherited Diseases
IncludesSystem only
OutputMax. Output Per Run (200 bp Reads): 15 Gb (1x540 Chip: 80M Reads/Chip), Max. Output Per Day (200 bp Reads): 15 Gb (1x540 Chip: 80M Reads/Chip)
Product TypeBenchtop Sequencing System
Quantity1 system
Sequencing Time200 bp: 2.5 hr. (500 Flows), 400 bp: 4 hr. (850 Flows), 600 bp: 4 hr. (1350 Flows)
Sequencing TypeBenchtop
Set Up Time (Hands-On Time)Library Prep on Chef: 15 min., Chip Loading on Chef: 15 min., Sequencing: 15 min.
Turn Around Time (Nucleic Acid to Variant Report)1 to 3 Days (Depending on the application)
Touch PointsLibrary Prep on Chef: 1, Chip Loading on Chef: 1, Sequencing: 1
Variant Calling and AnnotationIon Reporter Software
Weight63.5 kg
Software TypeTorrent Suite Software
Starting MaterialDNA, RNA
Unit SizeEach
Frequently asked questions (FAQs)
Can the Ion GeneStudio S5 System be used for diagnostic procedures?
The Ion GeneStudio S5 System (Cat. No. A38194) is labeled For Research Use Only and is not intended for use in diagnostic procedures. Diagnostic workflows require instruments, assays, and software appropriate for the intended diagnostic use. Contact Technical Support for product-specific guidance.
How many reads can I expect from an Ion 540 Chip on the Ion S5 System?
An Ion 540 Chip on the Ion S5 System typically generates approximately 60-80 million reads per run. Actual output can vary by library type, run quality, and application. For pricing or run-cost estimates, contact your sales representative or refer to current quote documentation.
What software is needed to operate and monitor runs on the Ion GeneStudio S5 System?
The Ion GeneStudio S5 System uses the Torrent Suite Software to plan, monitor, track, and analyze sequencing runs. The Torrent Suite Software is pre-installed on the system.
What Ion Sphere Particle (ISP) format is used with Ion Chef workflows?
The Ion Sphere Particles (ISPs) used depend on the chip type selected for the workflow. Each Ion Chef kit includes ISPs that are specific to the corresponding chip type to support template preparation.
What is the difference between the Ion S5 System and the Ion GeneStudio S5 System?
The Ion S5 System and the Ion GeneStudio S5 System have the same functionality and support the same consumables, chip types, and assays. The Ion S5 System was rebranded as the Ion GeneStudio S5 System, and only the GeneStudio S5 System is currently manufactured.