Poly-D-Lysine is a chemically synthesized extracellular matrix used to facilitate cell adhesion to tissue culture-treated plastic and glass surfaces. Poly-D-Lysine자세히 알아보기
Have Questions?
카탈로그 번호
수량
A3890401
100 mL
카탈로그 번호 A3890401
제품 가격(KRW)
186,000
온라인 행사
Ends: 31-Dec-2026
206,000
할인액 20,000 (10%)
Each
수량:
100 mL
제품 가격(KRW)
186,000
온라인 행사
Ends: 31-Dec-2026
206,000
할인액 20,000 (10%)
Each
Poly-D-Lysine is a chemically synthesized extracellular matrix used to facilitate cell adhesion to tissue culture-treated plastic and glass surfaces. Poly-D-Lysine contains polymers with a molecular weight of 50,000–150,000 Daltons, which makes this product ideal for neuronal culture applications. This ready-to-use product is provided as a liquid at a concentration of 0.1 mg/mL. When used on Nunc cell culture plastics with B-27 supplements or the B-27 Plus Neuronal Culture System, Poly-D-Lysine maintains cell viability and normal growth characteristics in multiple neuronal cell types.
Key features include: • Synthetic and animal origin-free • Sterile-filtered for cell culture applications • Validated with B-27 Supplement and B-27 Plus Neuronal Culture System when used with Nunc plastic and glass bottom dishes
For Research Use Only. Not for use in diagnostic procedures.
사양
세포 유형Mammalian Cells
농도0.1 mg/mL
배양 유형Mammalian Cell Culture
제품라인Gibco
순도 또는 품질 등급Cell Culture Grade
수량100 mL
유통 기한12 Months
배송 조건Approved for shipment on Wet Ice
소스Animal origin-free, Synthetic
분류Animal Origin-free
형태Liquid
제품 유형Extracellular Matrix
멸균Sterile
Unit SizeEach
구성 및 보관
Store in refrigerator (2–8°C).
자주 묻는 질문(FAQ)
What solution is Poly-D-Lysine (Cat. No. A3890401) delivered in?
Yes, Poly-D-Lysine can undergo one freeze-thaw cycle without impacting performance. Please note that this does not extend the shelf-life of the product.
I coated my plates with Poly-D-Lysine and I'm noticing that the cells are dying within a few days of plating the cells. How can I ensure cell survival?
It is possible that the plates were not adequately rinsed with water prior to plating the cells. Residual Poly-D-Lysine is toxic and may be the cause of cell death. It is recommended to rinse the plates with water three times before exposing the cells to the matrix.
I coated my plates with Poly-D-Lysine and I'm noticing that the cells are not attaching well. What can I do to prevent the cells from lifting off?
When coating the surface with Poly-D-Lysine, make sure that the entire surface is fully covered and that the appropriate volume of coating matrix is applied.