Collibri™ Stranded RNA Library Prep Kit for Illumina™ Systems
Invitrogen™

Collibri™ Stranded RNA Library Prep Kit for Illumina™ Systems

Kits are designed for next-generation sequencing (NGS) on Illumina systems and support whole-transcriptome sequencing (human, mouse, rat), and mRNA sequencing (compatible with all species) applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesIndex IncludedNo. of Reactions
A39003024RNA Prep, Depletion KitNo24 Reactions
A38994024RNA Prep Kit OnlyYes24 Reactions
A38994096RNA Prep Kit OnlyYes96 Reactions
A39003096RNA Prep, Depletion KitNo96 Reactions
A39005024RNA Prep, Depletion, UD IndexYes24 Reactions
A39005096RNA Prep, Depletion, UD IndexYes96 Reactions
A38996024RNA Prep and UD IndexYes24 Reactions
A38996096RNA Prep and UD IndexYes96 Reactions
Catalog number A39003024
Price (USD)
2,653.65
Online exclusive
3,015.00
Save 361.35
Each
Includes:
RNA Prep, Depletion Kit
Index Included:
No
No. of Reactions:
24 Reactions
Request bulk or custom format
Price (USD)
2,653.65
Online exclusive
3,015.00
Save 361.35
Each
The Invitrogen Collibri Stranded RNA Library Preparation kits are designed for next-generation sequencing (NGS) on Illumina systems. The kit supports whole-transcriptome sequencing (human, mouse, rat) and mRNA sequencing (compatible with all species) applications.

The Invitrogen Collibri Stranded RNA Library Preparation kits are designed for next-generation sequencing (NGS) on Illumina systems. The kit supports whole-transcriptome sequencing (human, mouse, rat) and mRNA sequencing (compatible with all species) applications.

The Collibri Stranded RNA Library Preparation Kit:

  • Includes all necessary components in a single kit* for a quick use
  • Total RNA converted to sequencing-ready library within 4.5 hours
  • >98% strand specificity, efficient detection of non-coding RNA and high sensitivity of differential gene expression
  • Provides in-process visual feedback for easier handling
  • Is suitable for conventional automation to increase throughput

The Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina Systems is designed for robust construction of cDNA libraries for strand-specific RNA sequencing on Illumina next-generation sequencing (NGS) systems. The kit brings together the superior features of SuperScript IV Reverse Transcriptase, Dynabeads magnetic particles, and Platinum SuperFi DNA Polymerase, enabling users to obtain high-quality sequencing-ready libraries. This Collibri Stranded RNA Library Prep Kit supports fast library preparation within 4.5 hours for mRNA or whole transcriptome libraries with 1–25 ng of mRNA-enriched or ribodepleted RNA sample input. Multiplexing of libraries can be carried out using up to 96 single-indexed or unique dual-indexed primers, enabling single-read or paired-end sequencing. The kit is suitable for various-quality RNA samples, including FFPE samples.

Visual cues of process integrity

For maximum convenience, visual cues of process integrity are included throughout the library preparation workflow. Monitor progress of library generation using visual feedback from dyes in critical reagents. The reaction mix changes color in response to the addition of critical components at each step to ensure that library preparation has the highest chance of success. The inert dyes do not interfere with enzymatic reactions and do not compromise sequencing results.

How it works

Full-length Illumina-compatible adaptors are introduced by PCR amplification using Platinum SuperFi DNA Polymerase to construct single-indexed or unique dual-indexed libraries compatible with single-read or paired-end sequencing. The kit contains 24 or 96 either single-indexed barcoded PCR primers (i7) pre-mixed with a universal i5 primer or unique dual-indexed PCR primers that enable multiplexing of up to 96 libraries. Optimized cleanup steps efficiently remove residual primers and adapter/primer dimers while preserving high library yields.

A related product, the Collibri Library Quantification Kit, is recommended for qPCR-based quantifications of libraries before proceeding to sequencing.

Applications

The Collibri Stranded RNA Library Prep Kit is recommended for:

  • Gene expression studies
  • Alternative splicing analysis
  • Non-coding RNA detection and discovery
  • Identification of alternative polyadenylation sites
  • Gene fusion detection
  • Detection of transcript isoforms

* The kit consists of enzymatic modification, size selection/clean-up, and indexed adapter-containing modules. Additional ribodepletion module of human/mouse/rat rRNA can also be included.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionThe product contains modules A39117024, A39118024, A39115024, A39100024
For Use With (Application)Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NextSeq 1000 and 2000, NovaSeq 6000
IncludesRNA Prep, Depletion Kit
Index IncludedNo
LibrariescDNA Libraries
No. of Reactions24 Reactions
Product LineCollibri, Illumina
Product TypeStranded RNA Library Prep Kit
Quantity24 preps
Sequencing TypeTranscriptome Sequencing
Shipping ConditionDry Ice
Workflow StepAdaptor Hybridization, Cleanup, Fragmentation, Indexing, Ligation, PCR Amplification, rRNA Depletion, Reverse Transcription
FormatKit
Sample TypeTotal RNA
SpeciesHuman, Mouse, Rat
Unit SizeEach
Contents & Storage
• Collibri Stranded RNA Library Prep Kit (24 preps). Store at -20°C.
• Collibri Library Cleanup Kit (24 preps). Store at 4°C.
• Collibri H/M/R rRNA Depletion Kit (24 preps). Store at 4°C.

Frequently asked questions (FAQs)

Is the elution buffer, included in the Collibri Stranded RNA Library Prep Kit for Illumina Systems, "Tris buffered" or "Tris-Tween"?

Collibri Stranded RNA Library Prep Kit for Illumina Systems elution buffer is based on Tris-HCl, with no EDTA or EGTA.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

Is there an RNA control for optimizing fragment length included with the Collibri Stranded RNA Library Prep Kit for Illumina Systems?

The Collibri Stranded RNA Library Prep Kit for Illumina Systems does not include an RNA control. Recommendations for RNA controls can be found in the Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina User Guide, page 16 "Guidelines for RNA Controls".

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What are the recommended RNA input sample types and respective sample amounts for Collibri Stranded RNA Library Prep Kit for Illumina Systems?

Information for RNA sample type and amount can be found in the Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina User Guide, page 15 "Guidelines for RNA sample type and amount".

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What are the differences between all Collibri Stranded RNA Library Prep Kit for Illumina Systems catalog numbers?

Information regarding the different catalog numbers can be found in the Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina User Guide, page 3.
The different catalog numbers allow for the option to purchase different sizes of the library kit and with or without Unique Dual Indexes.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What additional materials are required when using Collibri Stranded RNA Library Prep Kit for Illumina Systems?

A list of required materials that are not supplied with the kit can be found in the Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina User Guide, page 11-12.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

Citations & References (4)

Citations & References
Abstract
Transposon-associated TnpB is a programmable RNA-guided DNA endonuclease.
Authors:Karvelis T, Druteika G, Bigelyte G, Budre K, Zedaveinyte R, Silanskas A, Kazlauskas D, Venclovas C, Siksnys V
Journal:Nature
PubMed ID:34619744
'Transposition has a key role in reshaping genomes of all living organisms'
Evaluation of cisplatin-induced injury in human kidney organoids.
Authors:Digby JLM, Vanichapol T, Przepiorski A, Davidson AJ, Sander V
Journal:Am J Physiol Renal Physiol
PubMed ID:32150447
Acute kidney injury (AKI) remains a major global healthcare problem, and there is a need to develop human-based models to study AKI in vitro. Toward this goal, we have characterized induced pluripotent stem cell-derived human kidney organoids and their response to cisplatin, a chemotherapeutic drug that induces AKI and preferentially ... More
In situ approaches show the limitation of the spoilage potential of Juniperus phoenicea L. essential oil against cold-tolerant Pseudomonas fluorescens KM24.
Authors:Myszka K, Tomas N, Wolko L, Szwengiel A, Grygier A, Nuc K, Majcher M
Journal:Appl Microbiol Biotechnol
PubMed ID:33988734
The present study aimed to elucidate the effect of subinhibitory concentrations (sub-MICs) of juniper essential oil (EO), a-pinene, and sabinene on the quorum-sensing (QS)-mediated proteolytic and lipolytic properties of Pseudomonas fluorescens KM24. These activities were verified under in situ conditions, in which sub-MICs of the agents altered the morphology of ... More
TDP-43 prevents endogenous RNAs from triggering a lethal RIG-I-dependent interferon response.
Authors:Dunker W, Ye X, Zhao Y, Liu L, Richardson A, Karijolich J
Journal:Cell Rep
PubMed ID:33852834
RIG-I-like receptors (RLRs) are involved in the discrimination of self versus non-self via the recognition of double-stranded RNA (dsRNA). Emerging evidence suggests that immunostimulatory dsRNAs are ubiquitously expressed but are disrupted or sequestered by cellular RNA binding proteins (RBPs). TDP-43 is an RBP associated with multiple neurological disorders and is ... More