TaqMan™ Universal DNA Spike-In Control - FAQs
TaqMan™ Universal DNA Spike-In Control - FAQs
View additional product information for TaqMan™ Universal DNA Spike-In Control - FAQs (A39175)
2 product FAQs found
What is the size of TaqMan Universal DNA Spike-In Control (Cat. No. A39175)?
The TaqMan Universal DNA Spike-In Control (Cat. No. A39175) is a linearized plasmid less than 5 kb in size.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center
How can I check for the presence of inhibitors in my samples when running microbiota profiling experiments?
We offer a TaqMan Universal DNA Spike-In Control (Cat. No. A39175 or A39176) that can be added to samples before extraction, to monitor recovery efficiency.