What targets are in the TaqMan Respiratory Tract Microbiota Amplification Control?
The TaqMan Respiratory Tract Microbiota Amplification Control contains all the assays shown in Tables 1 and 2, page 7-8 and page 15 of the Respiratory Tract Microbiota Profiling Experiments v2 Application Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0019506_RespiratoryTractMicrobiotaProfiling_OAv2_AG.pdf).